Skip to Content
Merck
All Photos(1)

Key Documents

39326

Sigma-Aldrich

N-[2-(Dansylamino)ethyl]maleimide

BioReagent, suitable for fluorescence, ≥99.0% (HPLC)

Synonym(s):

N-(2-Maleimidoethyl)-5-dimethylamino-1-naphthalenesulfonamide, N-(2-Maleimidoethyl)dansylamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$215.00

$215.00


Estimated to ship onMay 22, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$215.00

About This Item

Empirical Formula (Hill Notation):
C18H19N3O4S
CAS Number:
Molecular Weight:
373.43
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.32

$215.00


Estimated to ship onMay 22, 2025


Request a Bulk Order

product line

BioReagent

Assay

≥99.0% (HPLC)

fluorescence

λex 327 nm; λem 547 nm in 0.1 M phosphate pH 7.0, 2-mercaptoethanol

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

CN(C)c1cccc2c(cccc12)S(=O)(=O)NCCN3C(=O)C=CC3=O

InChI

1S/C18H19N3O4S/c1-20(2)15-7-3-6-14-13(15)5-4-8-16(14)26(24,25)19-11-12-21-17(22)9-10-18(21)23/h3-10,19H,11-12H2,1-2H3

InChI key

GFVSIWWKMOVBJL-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Aoki et al.
Journal of the American Chemical Society, 123(6), 1123-1132 (2001-07-18)
A cyclen (=1,4,7,10-tetraazacyclododecane) doubly functionalized with three carbamoylmethyl groups and one dansylaminoethyl (dansyl = 2-(5-(dimethylamino)-1-naphthalenesulfonyl) group (L(2) = 1-(2-(5-(dimethylamino)-1-naphthalenesulfonylamido)ethyl)-4,7,10-tris(carbamoylmethyl)-cyclen) was synthesized and characterized. Potentiometrtic pH titration and UV spectrophotometric titration of L(2) served to determine deprotonation of the pendant dansylamide
Jun Yin et al.
Langmuir : the ACS journal of surfaces and colloids, 25(19), 11367-11374 (2009-08-28)
We report on the fabrication of Cu2+-sensing thermoresponsive poly(N-isopropylacrylamide) (PNIPAM) microgels labeled with metal-chelating acceptor and fluorescent reporter moieties. Cu2+ detection sensitivity can be considerably enhanced via thermo-induced collapse of the sensing matrix, which can easily optimize the relative spatial
Andreas Helmersson et al.
Plant physiology, 147(3), 1158-1167 (2008-05-30)
Zinc is a potent regulator of programmed cell death (PCD) in animals. While certain, cell-type-specific concentrations of intracellular free zinc are required to protect cells from death, zinc depletion commits cells to death in diverse systems. As in animals, PCD

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service