Skip to Content
Merck
All Photos(1)

Documents

36676

Supelco

Endosulfan sulfate

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6Cl6O4S
CAS Number:
Molecular Weight:
422.92
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

ClC1=C(Cl)[C@]2(Cl)C3COS(=O)(=O)OCC3[C@@]1(Cl)C2(Cl)Cl

InChI

1S/C9H6Cl6O4S/c10-5-6(11)8(13)4-2-19-20(16,17)18-1-3(4)7(5,12)9(8,14)15/h3-4H,1-2H2/t3?,4?,7-,8+

InChI key

AAPVQEMYVNZIOO-WINLOITPSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lili Niu et al.
Environmental pollution (Barking, Essex : 1987), 219, 315-322 (2016-11-05)
Organochlorine pesticides (OCPs) with different usage states, such as currently, historically or never used, may show different behaviors and potential risks in the environment. It is essential to identify their distribution patterns and fates and to assess their associated health
Reginald Quansah et al.
Environmental research, 150, 245-254 (2016-06-20)
Indiscriminate use of pesticides is a common practice amongst farmers in Low and Middle Income Countries (LMIC) across the globe. However, there is little evidence defining whether pesticide use is associated with respiratory symptoms. This cross-sectional study was conducted with
Thanh-Son Dao et al.
The Science of the total environment, 574, 1360-1370 (2016-08-17)
Metal contamination is one of the major issues to the environment worldwide, yet it is poorly known how exposure to metals affects tropical species. We assessed the sensitivity of a tropical micro-crustacean Daphnia lumholtzi to three trace metals: copper (Cu)
Anping Zhang et al.
The Science of the total environment, 505, 1142-1147 (2014-12-04)
The application of greenhouse vegetable cultivation has dramatically expanded worldwide during the last several decades. However, little information is available on the distribution and uptake of pesticides in greenhouse vegetables. To bridge this knowledge gap, the present study was initiated
Yunbin Jiang et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 983-984, 47-54 (2015-01-27)
A rapid and accurate multi-residue method was developed for simultaneously monitoring and determining 34 kinds of pesticides by GC-EI/MS/MS, successfully applied in Qianjinzhidai pills. Precision, repeatability, stability, limit of detection (LOD), limit of quantification (LOQ), linearity range and accuracy tests

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service