Merck
  • Distribution and uptake pathways of organochlorine pesticides in greenhouse and conventional vegetables.
Distribution and uptake pathways of organochlorine pesticides in greenhouse and conventional vegetables.
The Science of the total environment (2014-12-04)
Anping Zhang, Wenxiu Luo, Jianqiang Sun, Hang Xiao, Weiping Liu
ABSTRACT
The application of greenhouse vegetable cultivation has dramatically expanded worldwide during the last several decades. However, little information is available on the distribution and uptake of pesticides in greenhouse vegetables. To bridge this knowledge gap, the present study was initiated to investigate the distribution and uptake of organochlorine pesticides (OCPs) in vegetables from plastic greenhouse and conventional cultivation methods. The uptake pathways of OCPs were not significantly different between these two cultivation methods. The arithmetic means of OCP concentrations in greenhouse vegetables were higher than those in conventional vegetables, although there was no significant difference. This small difference raised the concern of whether the tiny difference could be magnified to a significant difference by bioaccumulation in the food chain. The issue should be addressed by a well-designed scheme in future studies.
Sigma-Aldrich
Pentachloronitrobenzene, ≥94%
Supelco
2,4′-DDD, PESTANAL®, analytical standard
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Supelco
Quintozene, PESTANAL®, analytical standard
Sigma-Aldrich
Hexane, suitable for HPLC, ≥95%
Sigma-Aldrich
Hexane, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
Hexane, Laboratory Reagent, ≥95%
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Hexane, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Supelco
Hexane, analytical standard
Sigma-Aldrich
Hexane, suitable for HPLC, ≥97.0% (GC)
Sigma-Aldrich
Hexane, ReagentPlus®, ≥99%