Merck
  • Development of a more specific and accurate multiple reaction monitoring method based on GC-EI/MS/MS for simultaneously monitoring and determining 34 kinds of pesticides in Qianjinzhidai pills.
Development of a more specific and accurate multiple reaction monitoring method based on GC-EI/MS/MS for simultaneously monitoring and determining 34 kinds of pesticides in Qianjinzhidai pills.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-01-27)
Yunbin Jiang, Mei Zhong, Yan Gou, Wei Peng, Juan Zhou, Pie Wu, Yuying Ma
ABSTRACT
A rapid and accurate multi-residue method was developed for simultaneously monitoring and determining 34 kinds of pesticides by GC-EI/MS/MS, successfully applied in Qianjinzhidai pills. Precision, repeatability, stability, limit of detection (LOD), limit of quantification (LOQ), linearity range and accuracy tests were used to validate this method. The results, precision (RSD<2%, n=6), repeatability (RSD<2%, n=6) and stability (RSD<2%, n=6), LOD (lower than 0.02mg/kg), LOQ (lower than 0.067mg/kg), linearity range (0.016-39.270μg/mL), recoveries (90-110%, RSD<15%, n=6), indicated that the method was feasible. This paper provided an optimization method about ion transitions which can improve the specificity of mass spectrum monitoring and the accuracy of content determination. This method can be also used to monitor and determine pesticide residues of other traditional Chinese medicine, but it was worth noting that the ion transitions should been again optimized according to the sample matrix.
Product Description
Supelco
Ethion, PESTANAL®, analytical standard
Supelco
Methanol, analytical standard
Supelco
Dimethoate, PESTANAL®, analytical standard
Supelco
Methidathion, PESTANAL®, analytical standard
Supelco
Acephate, PESTANAL®, analytical standard
Sigma-Aldrich
Pentachloronitrobenzene, ≥94%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Omethoate, PESTANAL®, analytical standard
Supelco
Quintozene, PESTANAL®, analytical standard
Supelco
Diazinon, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Methanol solution, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard