Skip to Content
Merck
All Photos(2)

Documents

1.06395

Supelco

Sodium carbonate

anhydrous 99.999 Suprapur®

Synonym(s):

anhydrous soda, Calcined soda, Carbonic acid disodium salt, Soda ash

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2CO3
CAS Number:
Molecular Weight:
105.99
Beilstein:
4154566
MDL number:
UNSPSC Code:
12352302
EC Index Number:
207-838-8
NACRES:
NA.21

grade

for inorganic trace analysis

Quality Level

Assay

99.999%

form

powder

potency

4090 mg/kg LD50, oral (Rat)

pH

11.16 (25 °C, 4 g/L in H2O)

mp

851 °C (lit.)

solubility

212.5 g/L

density

2.53 g/cm3 at 20 °C

bulk density

1100 kg/m3

storage temp.

2-30°C

SMILES string

O=C(O[Na])O[Na]

InChI

1S/CH2O3.2Na/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

CDBYLPFSWZWCQE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • A mixed separation-immobilization method for soluble salts removal and stabilization of heavy metals in municipal solid waste incineration fly ash.: Details a novel method using sodium carbonate for stabilizing heavy metals in waste ash, improving environmental safety and management of industrial by-products (Atanes et al., 2019).

Biochem/physiol Actions

Buffer component, may be used for the removal of peripheral membrane proteins.

Analysis Note

Purity (metallic): ≥ 99.999 %
Assay (acidimetric): ≥ 99.5 %
Chloride (Cl): ≤ 10 ppm
Phosphate (PO₄): ≤ 0.05 ppm
Silicate (SiO₂): ≤ 5 ppm
Sulfate (SO₄): ≤ 10 ppm
Al (Aluminium): ≤ 0.05 ppm
As (Arsenic): ≤ 0.1 ppm
Ba (Barium): ≤ 5.0 ppm
Ca (Calcium): ≤ 0.10 ppm
Cd (Cadmium): ≤ 0.005 ppm
Ce (Cerium): ≤ 0.010 ppm
Co (Cobalt): ≤ 0.005 ppm
Cr (Chromium): ≤ 0.010 ppm
Cu (Copper): ≤ 0.005 ppm
Eu (Europium): ≤ 0.010 ppm
Fe (Iron): ≤ 0.05 ppm
Hg (Mercury): ≤ 0.05 ppm
K (Potassium): ≤ 1.0 ppm
La (Lanthanum): ≤ 0.010 ppm
Li (Lithium): ≤ 0.5 ppm
Mg (Magnesium): ≤ 0.10 ppm
Mn (Manganese): ≤ 0.010 ppm
Ni (Nickel): ≤ 0.020 ppm
Pb (Lead): ≤ 0.010 ppm
Sc (Scandium): ≤ 0.010 ppm
Sm (Samarium): ≤ 0.010 ppm
Sr (Strontium): ≤ 0.10 ppm
Tl (Thallium): ≤ 0.01 ppm
Y (Yttrium): ≤ 0.010 ppm
Yb (Ytterbium): ≤ 0.010 ppm
Zn (Zinc): ≤ 0.010 ppm

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service