Skip to Content
Merck
All Photos(8)

Key Documents

795429

Sigma-Aldrich

Sodium hydroxide

anhydrous, free-flowing, pellets, Redi-Dri, ACS reagent, ≥97%

Synonym(s):

‘Caustic soda’

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$221.00
1 KG
$329.00
2.5 KG
$565.00
12 KG
$710.00

$221.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 G
$221.00
1 KG
$329.00
2.5 KG
$565.00
12 KG
$710.00

About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
EC Number:
MDL number:
UNSPSC Code:
12352316
PubChem Substance ID:
NACRES:
NA.21

$221.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

ACS reagent
anhydrous

Quality Level

Agency

suitable for DIN 38407-42
suitable for EPA 1621
suitable for EPA 533
suitable for EPA 8327
suitable for EPA OTM-45
suitable for SM 4500 - NH3

vapor density

>1 (vs air)

vapor pressure

<18 mmHg ( 20 °C)
3 mmHg ( 37 °C)

product line

Redi-Dri

Assay

≥97%

form

pellets

quality

free-flowing

impurities

≤0.001% N compounds
≤0.02% NH4OH ppt.
≤1.0% Na2CO3

mp

318 °C (lit.)

solubility

water: soluble 1260 g/L at 20 °C

density

2.13 g/cm3

anion traces

chloride (Cl-): ≤0.005%
phosphate (PO43-): ≤0.001%
sulfate (SO42-): ≤0.003%

cation traces

Fe: ≤0.001%
Hg: ≤0.1 ppm
K: ≤0.02%
Ni: ≤0.001%
heavy metals (as Ag): ≤0.002%

SMILES string

[OH-].[Na+]

application(s)

PFAS testing

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium hydroxide (NaOH) also known as caustic soda is a water soluble inorganic base with a wide range of industrial application such as titration, dissolution testing and in impinger to remove acidic gases. It participates in the oxidation of glycerol catalyzed by Au/charcoal or Au/graphite. It participates in solvent-free aldol condensation reaction.

Application

Sodium hydroxide (NaOH) was used in the following processes:
  • Preparation of sodium 3,3′-bis(sulfonato)-4,4′-bis(chloroacetamido)azobenzene (BSBCA) that is utilized in protein cross-linking techniques.
  • Preparation of electrolyte solution to study the effect of electrolyte composition on the conversion of CO2 to CO by electrochemical reduction.
  • Preparation of CSK (cytoskeleton) buffer for the three-dimensional (3D) slide method of preserving the 3D chromatin structure of testicular germ cells.
  • Synthesis of ZSMs (zeolite socony mobils).


Sodium hydroxide may be employed for the deterimination of specific surface area of colloidal silica, via titration. It may be employed for the direct oxidation of sodium borohydride, which was investigated by cyclic and linear voltammetry, chronoamperometry and chronopotentiometry. It may be used to prepare citrate buffer and borate buffer.

It may be used in the synthesis of :
  • alkylaryl amidosulfobetaines
  • (3 -[4-teroctylphenoxyethoxyethoxyethyl] dimethylammonio propane sulfonate (XOSB)
  • trisulfobetaines (TriSBn)


Sodium hydroxide may be used as a base in titration methods, dissolution testing, for adjusting pH and in the impinger to remove acidic gases.

It may be used in the following processes:
  • Conversion of glycerol to glyceric acid in the presence of gold catalyst.
  • Palladium-catalyzed cross-coupling reaction between vinylalkoxysilanes and aryl bromides or chlorides to form styrenes.

Features and Benefits

  • Increased efficiency – Eliminate time and effort of loosening hardened powders
  • Safety – Avoid risks of using tools of force to break up clumps
  • Economy – Faster preparation and solvation increase productivity and reduce costs
  • Assured quality – Excellent, expert-tested quality with no anti-caking agents
  • Flexible volumes – Available from research amounts to scale-up quantities

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bilal Çakir et al.
Plant foods for human nutrition (Dordrecht, Netherlands), 74(3), 414-420 (2019-07-07)
The amount of cold press oil manufacture is globally rising, which in turn leads to the accumulation of deoiled plant seeds at significant quantities and consequent manufacture of plant protein products. In this study, we made an attempt to analyze
You Fu et al.
International journal of molecular medicine, 46(4), 1501-1513 (2020-07-24)
The lack of reliable animal models to assess the safety and efficacy of drugs and to explore the underlying molecular mechanisms is one of the most severe impediments in head and neck squamous cell carcinoma (HNSCC) tumor immunology research. The
Hanyu Zhang et al.
Nanoscale, 12(15), 8344-8354 (2020-04-03)
By understanding how the environmental composition impacts the optoelectronic properties of transition metal dichalcogenide monolayers, we demonstrate that simple photoluminescence (PL) measurements of tungsten disulfide (WS2) monolayers can differentiate relative humidity environments. In this paper, we examine the PL and
Debonil Maity et al.
Soft matter, 15(12), 2617-2626 (2019-02-28)
Extracellular matrices in animal tissue are hydrogels mostly made of collagen. In these matrices, collagen fibers are hierarchically assembled and cross-linked to form a porous and elastic material, through which migrating cells can move by either pushing through open matrix
Ruobing Bai et al.
Macromolecular rapid communications, 40(8), e1800883-e1800883 (2019-02-12)
New applications of hydrogels draw growing attention to the development of tough hydrogels. Most tough hydrogels are designed through incorporating large energy dissipation from breaking sacrificial bonds. However, these hydrogels still fracture under prolonged cyclic loads with the presence of

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service