Skip to Content
Merck
All Photos(9)

Key Documents

746398

Sigma-Aldrich

Sodium chloride

anhydrous, Redi-Dri, free-flowing, ACS reagent, ≥99%

Synonym(s):

Halite, NaCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
anhydrous
form:
powder
Pricing and availability is not currently available.

grade

ACS reagent
anhydrous

Quality Level

Agency

suitable for GB 5009.253-2016

vapor pressure

1.33 hPa ( 865 °C)

product line

Redi-Dri

Assay

≥99%

form

powder

quality

free-flowing

impurities

≤0.005% Insoluble matter

pH

5.0-9.0 (25 °C, 5% in solution)

mp

801 °C (lit.)

anion traces

bromide (Br-): ≤0.01%
chlorate, nitrate (as NO3-): ≤0.003%
iodide (I-): ≤0.002%
phosphate (PO43-): ≤5 ppm
sulfate (SO42-): ≤0.004%

cation traces

Ba:, passes test
Ca: ≤0.002%
Fe: ≤2 ppm
K: ≤0.005%
Mg: ≤0.001%
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Redi-Dri technology eliminates clumping and the need for complicated procedures to keep your salts free-flowing. Keeping your salts dry helps improve your results by giving accurate measurements and easier-to-handle salts and buffers. Rather than drying your salts in an oven or desiccator or breaking them apart manually, Redi-Dri ensures that your salts will remain easy-to-use for months without worrying about anti-caking agents. Even after repeated openings you can be assured that your results are always accurate. Redi-Dri saves you time, energy and money by keeping your salts usable for longer.

Application

Sodium chloride has been used for the preparation of tris buffered saline, phosphate buffered saline, MPM-2 (mitotic protein monoclonal-2) cell lysis buffer, immunoprecipitation wash buffer, LB (Luria-Bertani) media and dialysis buffer.

Features and Benefits

  • Increased efficiency – Eliminate time and effort of loosening hardened powders
  • Safety – Avoid risks of using tools of force to break up clumps
  • Economy – Faster preparation and solvation increase productivity and reduce costs
  • Assured quality – Excellent, expert-tested quality with no anti-caking agents
  • Flexible volumes – Available from research amounts to scale-up quantities

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aaron S Schwartz-Duval et al.
Nature communications, 11(1), 4530-4530 (2020-09-12)
Various cancer cells have been demonstrated to have the capacity to form plasmonic gold nanoparticles when chloroauric acid is introduced to their cellular microenvironment. But their biomedical applications are limited, particularly considering the millimolar concentrations and longer incubation period of
Jinbo Cheng et al.
Autophagy, 16(12), 2193-2205 (2020-02-01)
Microglial activation-induced neuroinflammation is closely associated with the development of Parkinson disease (PD). Macroautophagy/autophagy regulates many biological processes, but the role of autophagy in microglial activation during PD development remains largely unclear. In this study, we showed that deletion of
Anneleen Remmerie et al.
Immunity, 53(3), 641-657 (2020-09-06)
Metabolic-associated fatty liver disease (MAFLD) represents a spectrum of disease states ranging from simple steatosis to non-alcoholic steatohepatitis (NASH). Hepatic macrophages, specifically Kupffer cells (KCs), are suggested to play important roles in the pathogenesis of MAFLD through their activation, although
Islem Gandouzi et al.
Materials (Basel, Switzerland), 12(7) (2019-04-14)
Pyoverdine is a fluorescent siderophore produced by Pseudomonas aeruginosa that can be considered as a detectable marker in nosocomial infections. The presence of pyoverdine in water can be directly linked to the presence of the P. aeruginosa, thus being a
Johnny Bonnardel et al.
Immunity, 51(4), 638-654 (2019-09-29)
Macrophages are strongly adapted to their tissue of residence. Yet, little is known about the cell-cell interactions that imprint the tissue-specific identities of macrophages in their respective niches. Using conditional depletion of liver Kupffer cells, we traced the developmental stages

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Protocols

Overcoated SPME fiber enables low-level BPA analysis in food via SPME-GC-MS/MS, meeting tighter migration limit regulations.

Questions

1–5 of 5 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. How long will material remain free-flowing after it is opened?

    1 answer
    1. While the exact amount of time your material will remain free-flowing can vary, this largely depends on the ambient storage conditions and how many times it is opened.

      Helpful?

  4. How does Redi-Dri™ help keep salts and buffers free-flowing?

    1 answer
    1. Redi-Dri™ products are specially handled and the bottle is specially designed to ensure that the product will remain dry without adding impurities or additives to the product. Ensuring that you receive the highest quality material that is easy to handle.

      Helpful?

  5. Are there any special storage conditions that must be maintained?

    1 answer
    1. No, Redi-Dri™ products can be stored under the same conditions as the equivalent non-Redi-Dri™ products.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service