Skip to Content
Merck
Sodium hydroxide anhydrous, free-flowing, Redi-Dri™, reagent grade, ≥98%, pellets

Sodium hydroxide

Sodium hydroxide anhydrous, free-flowing, Redi-Dri™, reagent grade, ≥98%, pellets
Synonym(s):
‘Caustic soda’, Sodium hydroxide
Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade
Technique
Showing 1-20 of 20
Compare
Product Number
Description
Pricing