Skip to Content
Merck
All Photos(1)

Key Documents

60349

Sigma-Aldrich

Potassium phosphate dibasic trihydrate

puriss. p.a., ≥99.0% (T)

Synonym(s):

Dipotassium hydrogen phosphate trihydrate, Dipotassium phosphate, di-Potassium hydrogen phosphate trihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$206.00

$206.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 G
$206.00

About This Item

Linear Formula:
K2HPO4 · 3H2O
CAS Number:
Molecular Weight:
228.22
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
form:
crystals

$206.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

puriss. p.a.

Assay

≥99.0% (T)

form

crystals

impurities

≤0.001% total nitrogen (N)

pH

9.1-9.4 (25 °C, 50 mg/mL in H2O)

anion traces

chloride (Cl-): ≤20 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

As: ≤0.1 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤500 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

O.O.O.[K+].[K+].OP([O-])([O-])=O

InChI

1S/2K.H3O4P.3H2O/c;;1-5(2,3)4;;;/h;;(H3,1,2,3,4);3*1H2/q2*+1;;;;/p-2

InChI key

XQGPKZUNMMFTAL-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Potassium phosphate dibasic trihydrate (K2HPO4.3H2O, Dipotassium hydrogen phosphate trihydrate, Dipotassium phosphate, di-Potassium hydrogen phosphate trihydrate) is the potassium salt having a pH in the range of 9.1-9.4.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lewis Stevens et al.
Current biology : CB, 30(6), 1023-1031 (2020-02-29)
The free-living nematode Caenorhabditis elegans is a key laboratory model for metazoan biology. C. elegans has also become a model for parasitic nematodes despite being only distantly related to most parasitic species. All of the ∼65 Caenorhabditis species currently in culture are
L Turner et al.
Biophysical journal, 77(1), 597-603 (1999-07-02)
The behavior of the bacterium Escherichia coli is controlled by switching of the flagellar rotary motor between the two rotational states, clockwise (CW) and counterclockwise (CCW). The molecular mechanism for switching remains unknown, but binding of the response regulator CheY-P
Silvio D Brugger et al.
Journal of clinical microbiology, 47(6), 1750-1756 (2009-04-24)
Colonization with more than one distinct strain of the same species, also termed cocolonization, is a prerequisite for horizontal gene transfer between pneumococcal strains that may lead to change of the capsular serotype. Capsule switch has become an important issue
Erfan Kalantari et al.
Materials science & engineering. C, Materials for biological applications, 98, 1087-1096 (2019-03-01)
In this research, novel monticellite/hydroxyapatite (HA) ceramic composites were successfully prepared by mechanical method. The ability of nanostructured monticellite-based ceramic composites to form a suitable bond to living hard tissues, and stimulate osteoblast-like cells proliferation may be different for various
Amperometric glucose biosensor based on NiFe2O4 nanoparticles and chitosan.
Luo L, et al.
Sensors and Actuators B, Chemical, 145(1), 293-298 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service