Skip to Content
Merck
All Photos(2)

Key Documents

259691

Sigma-Aldrich

Methylcyclohexane

spectrophotometric grade, 99%

Synonym(s):

Hexahydrotoluene

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$295.00
2 L
$496.00

$295.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
1 L
$295.00
2 L
$496.00

About This Item

Linear Formula:
C6H11CH3
CAS Number:
Molecular Weight:
98.19
Beilstein:
505972
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.21
grade:
spectrophotometric grade
technique(s):
UV/Vis spectroscopy: suitable
bp:
101 °C (lit.)
vapor pressure:
37 mmHg ( 20 °C)
83.2 mmHg ( 37.7 °C)

$295.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

grade

spectrophotometric grade

vapor density

3.4 (vs air)

vapor pressure

37 mmHg ( 20 °C)
83.2 mmHg ( 37.7 °C)

Assay

99%

form

liquid

autoignition temp.

545 °F

expl. lim.

6.7 %

technique(s)

UV/Vis spectroscopy: suitable

impurities

<0.010% water

evapn. residue

<0.0005%

refractive index

n20/D 1.422 (lit.)

bp

101 °C (lit.)

mp

−126 °C (lit.)

density

0.77 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 207 nm Amax: 1.00
λ: 221 nm Amax: 0.40
λ: 232 nm Amax: 0.15
λ: 260 nm Amax: 0.05
λ: 300-400 nm Amax: 0.01

SMILES string

CC1CCCCC1

InChI

1S/C7H14/c1-7-5-3-2-4-6-7/h7H,2-6H2,1H3

InChI key

UAEPNZWRGJTJPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methylcyclohexane is an organic compound that belongs to the class of saturated hydrocarbons. It is primarily used as a solvent in organic synthesis.

Application

Methylcyclohexane can be used as a model compound in fluid catalytic cracking reactions.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Internal conversion of o-aminoacetophenone in solution.
Yoshihara T, et al.
Physical Chemistry Chemical Physics, 3(22), 4972-4978 (2001)
Methylcyclohexane and methylcyclohexene cracking over zeolite Y catalysts.
Corma A, et al.
Applied Catalysis, 67(1), 307-324 (1990)
Eagleson M.
Concise Encyclopedia Chemistry, 293-293 (1994)
Liliana Martelo et al.
The journal of physical chemistry. B, 119(48), 15023-15029 (2015-11-10)
Phasor plots of the fluorescence intensity decay (plots of the Fourier sine transform versus the Fourier cosine transform, for one or several angular frequencies) are being increasingly used in studies of homogeneous and heterogeneous systems. In this work, the phasor
Brian D Fitz et al.
Journal of chromatography. A, 1392, 82-90 (2015-03-31)
Low thermal mass gas chromatography (LTM-GC) was evaluated for rapid, high peak capacity separations with three injection methods: liquid, headspace solid phase micro-extraction (HS-SPME), and direct vapor. An Agilent LTM equipped with a short microbore capillary column was operated at

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service