Skip to Content
Merck
All Photos(2)

Key Documents

153087

Sigma-Aldrich

2-Methylcyclohexanol, mixture of cis and trans

99%

Synonym(s):

Hexahydro-o-cresol

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$132.00
1 L
$367.00

$132.00


Estimated to ship onApril 02, 2025


Request a Bulk Order

Select a Size

Change View
250 ML
$132.00
1 L
$367.00

About This Item

Linear Formula:
CH3C6H10OH
CAS Number:
Molecular Weight:
114.19
Beilstein:
1847535
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$132.00


Estimated to ship onApril 02, 2025


Request a Bulk Order

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.462 (lit.)

bp

163-166 °C (lit.)

density

0.93 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CC1CCCCC1O

InChI

1S/C7H14O/c1-6-4-2-3-5-7(6)8/h6-8H,2-5H2,1H3

InChI key

NDVWOBYBJYUSMF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Methylcyclohexanol was used to study the effect of organic solvents on epoxide hydrolase[1].

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeanette Lotter et al.
Biotechnology letters, 26(15), 1191-1195 (2004-08-04)
Epoxides are often highly hydrophobic substrates and the presence of an organic co-solvent within an aqueous bioreactor is in such cases indicated. The effect of 40 water-miscible and -immiscible organic solvents on epoxide hydrolase activity in whole-cells of the yeast
Halis T Balaydın et al.
Bioorganic & medicinal chemistry letters, 22(3), 1352-1357 (2012-01-11)
The Naturally occurring novel cyclohexanonyl bromophenol 2(R)-2-(2,3,6-tribromo-4,5-dihydroxybenzyl)cyclohexanone (4) was synthesized as a racemic compound. Cyclohexylphenyl methane derivatives (10-17) with Br, OMe, CO, and OH were also obtained. Inhibition of four human carbonic anhydrase (hCA, EC 4.2.1.1) isozymes I, II, IV

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service