Skip to Content
Merck
All Photos(3)

Key Documents

243663

Sigma-Aldrich

Sodium hypophosphite hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$72.00
500 G
$200.00

$72.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$72.00
500 G
$200.00

About This Item

Linear Formula:
NaH2PO2 · xH2O
CAS Number:
Molecular Weight:
87.98 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.55

$72.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

~95% (NaH2PO2)

form

powder, crystals or chunks

SMILES string

O.[Na+].OP[O-]

InChI

1S/Na.H3O2P.H2O/c;1-3-2;/h;3H2,(H,1,2);1H2/q+1;;/p-1

InChI key

PLZNPHDJGFDNRM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Sodium hypophosphite hydrate can be used as:
  • A radical precursor to synthesize mono alkyl phosphinic acids via radical reaction with alkenes in the presence of Et3B as the initiator.[1]
  • A reagent for the reduction of the diazonium salt.[2]
  • A hydrogen donor in the transfer-hydrogenation reactions.[3]
  • A catalyst in the esterification reaction.[4]

It can also be used to compose the finishing solutions for the immobilization of photo-initiator (benzophenone ) onto cotton fabrics.[5]
Sodium hypophosphite hydrate is widely used as an esterification catalyst.[6] It may be used to compose the finishing solutions for the incorporation of benzophenone (photoinitiator) onto cotton fabrics.[5][7]

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of wood filler treatment and EBAGMA compatibilizer on morphology and mechanical properties of low density polyethylene/olive husk flour composites.
Kaci M, et al.
Express Polymer Letters, 1(7), 467-473 (2007)
Convenient synthesis of 4-methylhistamine and racemic. alpha., 4-dimethylhistamine and. alpha., 4-dimethylhistidine
Davey DD and Lumma Jr WC
The Journal of Organic Chemistry, 54(13), 3211-3213 (1989)
S Deprèle et al.
The Journal of organic chemistry, 66(20), 6745-6755 (2001-10-02)
A novel and practical approach to monosubstituted phosphinic acid (alkylphosphonous acid) derivatives from hypophosphite salts or esters is described. Phosphorus-centered radical formation is initiated with Et(3)B/O(2), and the reaction is conveniently conducted at room temperature in an open flask. In
X-Ray Photoelectron Spectroscopy of Lignocellulosic Materials Treated with Malea Ted Polypropylenes
Kazayawoko M, et al.
J. Wood Chem. Technol., 18(1), 1-26 (1998)
UV-induced graft polymerization of acrylamide on cellulose by using immobilized benzophenone as a photo-initiator.
Hong KH, et al.
European Polymer Journal, 45(8), 2443-2449 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service