Skip to Content
Merck
All Photos(3)

Documents

1.07209

Supelco

Hydrogen peroxide 30%

(Perhydrol) for analysis EMSURE ISO

Synonym(s):

Hydrogen peroxide solution, Perhydrol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H2O2
CAS Number:
Molecular Weight:
34.01
Beilstein:
3587191
MDL number:
UNSPSC Code:
12352304
NACRES:
NA.21

Agency

reag. ISO

Quality Level

vapor pressure

18 hPa ( 20 °C)
23.3 mmHg ( 30 °C)

product line

EMSURE®

pH

≤3.5 (20 °C in H2O)

bp

107 °C/1013 hPa

mp

-25.7 °C

density

1.11 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

OO

InChI

1S/H2O2/c1-2/h1-2H

InChI key

MHAJPDPJQMAIIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Hydrogen peroxide 30% EMSURE® grade product) is used for regulated analyses and highly demanding lab applications. It also offers the highest quality and unmatched scope of specifications to give complete control of test conditions and eliminate uncertainties. EMSURE® grade products comply with all international regulations and can be used in various applications.
Benefits of using EMSURE® grade products.
  • Most extensive specifications worldwide
  • Tested for up to 70 parameters
  • Extraordinary purity
  • Very low limiting values
  • Greater accuracy and control of analyses
  • Optimized for highly critical and demanding analyses
  • Ideal for method development
  • No interference or contamination due to unknown impurities

Application

Hydrogen peroxide 30% EMSURE®, ISO) can be used in the analysis of mercury from blood samples using a Perkin Elmer, FIMS 100 (flow injection mercury system). It can also be used along with sulfuric acid (piranha solution) to remove organic and inorganic contaminants from the substrate.

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany
PERHYDROL is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carmen Sarcinelli et al.
Cancers, 12(3) (2020-03-04)
Endoplasmic reticulum (ER) stress generates reactive oxygen species (ROS) that induce apoptosis if left unabated. To limit oxidative insults, the ER stress PKR-like endoplasmic reticulum Kinase (PERK) has been reported to phosphorylate and activate nuclear factor erythroid 2-related factor 2

Protocols

Preparation of a standard solution for Hydrogen peroxide

Photometric determination with PAR subsequent to acid mineralisation; DIN decomposition

Photometric determination with Cuprizon subsequent to acid mineralisation

Photometric determination with Dimethylglyoxime subsequent to acid mineralisation; DIN decomposition

See All

Related Content

Reflectometric peroxide testing in food packaging materials with RQflex® 20 reflectometer and Reflectoquant® Test Strips.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service