04283
Sodium phosphite dibasic pentahydrate
≥98%
Synonym(s):
di-Sodium hydrogen phosphite
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
Na2HPO3 · 5H2O
CAS Number:
Molecular Weight:
216.04
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22
grade:
for analytical purposes
form:
crystals
Recommended Products
grade
for analytical purposes
Quality Level
Assay
≥98%
form
crystals
impurities
≤0.001% heavy metals (as Pb)
anion traces
chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg
cation traces
Fe: ≤10 mg/kg
SMILES string
O.O.O.O.O.[Na+].[Na+].OP([O-])[O-]
InChI
1S/2Na.HO3P.5H2O/c;;1-4(2)3;;;;;/h;;1H;5*1H2/q2*+1;-2;;;;;
InChI key
FBKBMXXJECMLCQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Sodium phosphite dibasic pentahydrate can be used as an electron donor for the enantioselective transformation of prochiral sulfides to the corresponding optically active sulfoxides catalyzed by Thermocrispum municipale cyclohexanone monooxygenase (TmCHMO) enzyme.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Enantioselective sulfoxidations employing the thermostable cyclohexanone monooxygenase from Thermocrispum municipale.
De G G and Franconetti A
Enzyme and Microbial Technology, 113, 24-28 (2018)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service