Skip to Content
Merck
All Photos(1)

Key Documents

SMB01329

Sigma-Aldrich

Val-Pro-Pro TFA salt

Synonym(s):

VPP TFA salt, Valine-Proline-Proline TFA salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$94.50
25 MG
$379.00

$94.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$94.50
25 MG
$379.00

About This Item

Empirical Formula (Hill Notation):
C15H25N3O4 · xC2HF3O2
Molecular Weight:
311.38 (free base basis)
UNSPSC Code:
12352209
NACRES:
NA.26

$94.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98% (HPLC)

Quality Level

form

powder

color

white to off-white

storage temp.

−20°C

General description

Val-Pro-Pro TFA salt, a tripeptide commonly derived from milk caseins and composed of valine, proline, and proline, is a water-soluble salt that shows immense potential in biomedical and biochemical research. One of its key attributes is its role as a potent angiotensin-converting enzyme (ACE) inhibitor, a critical enzyme in the regulation of blood pressure[1]. Furthermore, Val-Pro-Pro TFA salt stimulates the production of nitric oxide (NO), a vasodilator responsible for relaxing blood vessels, which can contribute to its vasodilatory effects and potential in blood pressure management[2][3]. In addition to its cardiovascular effects, Val-Pro-Pro TFA salt is recognized for its anti-inflammatory and antioxidant properties, making it a promising candidate for biochemical and metabolomics research.

Application

Val-Pro-Pro TFA salt can be used in biochemical and metabolomics research applications

Features and Benefits

  • Suitable for Metabolomics and Biochemical research
  • High-quality compound suitable for multiple research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Radiology of the parapharyngeal space
Chong et al.
Australasian Radiology, 42, 278-283 (1998)
Renal transplantation and graft survival and its influencing factors
Agraz et al.
Transplantation Proceedings, 31, 296-300 (1999)
An evaluation of operator preference of diamond burs in coronal tooth preparation
Cohen BD et al.
Compendium of Continuing Education in Dentistry. (Jamesburg, N.J. : 1995). Supplement, 18, 158-162 (1997)
C Adams et al.
Journal of dairy science, 103(7), 5805-5815 (2020-05-26)
Lactic acid bacteria (LAB) are used as starter cultures in the production of fermented dairy products and have the potential to confer bioactivity relevant to cardiovascular health, as they possess extensive proteolytic systems that liberate small bioactive peptides from larger
Tianyuan Song et al.
Journal of agricultural and food chemistry, 68(47), 13730-13741 (2020-11-13)
Endothelial dysfunction is a common disorder of vascular homeostasis in hypertension characterized by oxidative stress, malignant migration, inflammatory response, and active adhesion response of endothelial cells. The extracellular vesicles (EVs), a vital participant in vascular cell communication, have been considered

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service