Skip to Content
Merck
All Photos(1)

Key Documents

R9032

Sigma-Aldrich

R-715 TFA salt

≥95% (HPLC)

Synonym(s):

Ac-Lys-[D-betaNal7,lle8]desArg9-bradykinin

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$257.00

$257.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
1 MG
$257.00

About This Item

Empirical Formula (Hill Notation):
C57H81N13O12 · xC2HF3O2
Molecular Weight:
1140.33 (free base basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$257.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Assay

≥95% (HPLC)

form

powder

color

white to off-white

solubility

H2O: >1 mg/mL

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.CC[C@H](C)[C@H](NC(=O)[C@@H](Cc1ccc2ccccc2c1)NC(=O)[C@H](CO)NC(=O)[C@H](Cc3ccccc3)NC(=O)CNC(=O)[C@@H]4CCCN4C(=O)[C@@H]5CCCN5C(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCCCN)NC(C)=O)C(O)=O

InChI

1S/C57H81N13O12.C2HF3O2/c1-4-34(2)48(56(81)82)68-51(76)43(31-37-23-24-38-17-8-9-18-39(38)29-37)66-52(77)44(33-71)67-50(75)42(30-36-15-6-5-7-16-36)64-47(73)32-62-53(78)45-21-13-27-69(45)55(80)46-22-14-28-70(46)54(79)41(20-12-26-61-57(59)60)65-49(74)40(63-35(3)72)19-10-11-25-58;3-2(4,5)1(6)7/h5-9,15-18,23-24,29,34,40-46,48,71H,4,10-14,19-22,25-28,30-33,58H2,1-3H3,(H,62,78)(H,63,72)(H,64,73)(H,65,74)(H,66,77)(H,67,75)(H,68,76)(H,81,82)(H4,59,60,61);(H,6,7)/t34-,40-,41-,42-,43+,44-,45-,46-,48-;/m0./s1

InChI key

ZFUJDXXJCMFOSK-WBUMRSSGSA-N

Amino Acid Sequence

Ac-Lys-Arg-Pro-Pro-Gly-Phe-Ser-Ile

Biochem/physiol Actions

Novel, selective, metabolically stable B1 bradykinin receptor antagonist.

Features and Benefits

This compound is featured on the Bradykinin Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service