Skip to Content
Merck
All Photos(3)

Key Documents

PPB001

Sigma-Aldrich

Tris Buffered Saline

powder, pH 8.0, pHast Pack

Synonym(s):

1X TBS wash buffer, TBS

Sign Into View Organizational & Contract Pricing

Select a Size

20 PKG
$131.00

$131.00


Estimated to ship onMarch 13, 2025


Request a Bulk Order

Select a Size

Change View
20 PKG
$131.00

About This Item

MDL number:
UNSPSC Code:
12161700
NACRES:
NA.21

$131.00


Estimated to ship onMarch 13, 2025


Request a Bulk Order

product name

Tris Buffered Saline, pH 8.0, pHast Pack, powder

product line

pHast Pack

form

powder

storage condition

(Tightly closed. Dry)

impurities

≤5 ppm Heavy Metals (lead)

color

white

pH

8.0

solubility

soluble, clear, colorless

cation traces

Fe: <10 ppm

foreign activity

DNAse, none detected
Nickase, none detected
Protease, none detected
RNAse, none detected

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Tris Buffered Saline, pH 8.0 is a commonly used wash buffer that is isotonic and nontoxic and suitable for use in cell culture and electrophoresis applications.

Application

Tris-buffered saline is intended to use for:
  • Sample preparation
  • ELISA
  • Western Blot and other protein applications
  • Cell culture (recommend to sterile filter)
  • Immunohistochemistry
  • Resuspension and sonication of cells

Features and Benefits

  • Ready-to-use buffer – fast and easy to prep
  • No need to measure or adjust pH, simply add water
  • Saves time and minimizes the effort
  • Biological tests: free of DNase, RNase, protease, and nickase
  • Chemical tests: Iron ≤10 ppm, lead ≤5 ppm

Packaging

Foil pouches

Reconstitution

Contents of one pouch, when dissolved in 500mL of distilled or deionized water, will yield a 1X solution containing 50mM Tris, 138mM NaCl, 2.7mM KCl, pH 8.0 at 25 °C. Certified to be DNAse, RNAse, Protease, and Nickase free. Certified for use in cell culture.

Storage and Stability

Store at room temperature. Product may naturally agglomerate but can be simply broken up within the pouch prior to use.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

pHast Pack is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service