Skip to Content
Merck
All Photos(1)

Key Documents

PHR2892

Supelco

Polyethylene Glycol 4000

certified reference material, pharmaceutical secondary standard

Synonym(s):

Poly(ethylene glycol), PEG

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$187.00

$187.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$187.00

About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.24

$187.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1546569

API family

polyethylene glycol

CofA

current certificate can be downloaded

packaging

pkg of 1 g

application(s)

pharmaceutical

storage temp.

2-30°C

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. Is this product suitable for quantitative analysis? Is the %purity of PEG4000 available on the CoA? How can you get the %purity?

    1 answer
    1. es, this product is suitable for quantitative analysis, such as content or assay tests. The % purity is available on the Certificate of Analysis. The purity is calculated using the following equation:
      % purity = (100 - ROI - LOD - H2O - RS) x ((100 - TCI)/100)
      Where:
      TCI = Total Chromatographic Impurities
      LOD = Loss on Drying
      H2O = Water content determined by Karl Fischer analysis
      ROI = Residue on Ignition
      RS = Residual Solvents

      Helpful?

  2. I ordered this material to use as a certified reference material for GPC determination. The certificate of analysis has a purity but not an average Mw, Mn or Mp. Can I please get the Mp for this standard so that I can use it for my purpose?

    1 answer
    1. This product is intended for use as a USP, EP, and BP traceable secondary standard in compendial methods. The testing performed is for purity by HPLC, moisture determination, and residue analysis. The agency specific monographs do not include GPC analysis or specifications for Mn, Mp, and Mw. Please see the link below to review a sample of lot specific Certificate of Analysis for a product that may be more suitable for GPC analysis.
      https://www.sigmaaldrich.com/certificates/Graphics/COfAInfo/fluka/pdf/rtc/PHR2892_LRAD7319.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service