Skip to Content
Merck
All Photos(1)

Key Documents

PHR2362

Supelco

Polyethylene Glycol 3350

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Poly(ethylene glycol), PEG

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$171.00

$171.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$171.00

About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$171.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1546547

API family

polyethylene glycol

form

solid

CofA

current certificate can be downloaded

packaging

pkg of 1 g

application(s)

pharmaceutical small molecule

storage temp.

2-30°C

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC4029 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Estamos interesados en comprar polietilenglicol PHR2362 PS y veo que es trazable al de USP 1546547: Polietilenglicol de peso molecular Mw 3350 (3015-3685) pero en el vuestro PHR2362 no se especifica el peso molecular. ¿Me lo podéis pasar? ¿Y un CoA?

    1 answer
    1. The '3350' designated in the name of this product represents the molecular weight. In addition, the molecular weight is published in the product Certificate of Analysis as approximately 3350 daltons. Please see the link below to review a sample or lot-specific Certificate of Analysis.
      https://www.sigmaaldrich.com/documents-search?tab=coa

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service