Skip to Content
Merck
All Photos(1)

Documents

81292

Supelco

Poly(ethylene glycol)

analytical standard, for GPC, 4,000

Synonym(s):

PEG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard
for GPC

Quality Level

shelf life

limited shelf life, expiry date on the label

mol wt

Mn ~4120
Mp ~4450
Mw ~4240

technique(s)

gel permeation chromatography (GPC): suitable

Mw/Mn

~1.03

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

C(CO)O

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(ethylene glycol) (PEG) is regarded as the gold standard for stealth polymers in polymer based drug delivery. PEGs and their anionic or non-ionic derivatives are widely used in cosmetic products.

Application

PEG is known to modify enzymes and proteins for the preparation of improved therapeutic agents.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Safety assessment on polyethylene glycols (PEGs) and their derivatives as used in cosmetic products.
Fruijtier-Polloth C.
Toxicology, 214(1-2), 1- 38 (2005)
C Woghiren et al.
Bioconjugate chemistry, 4(5), 314-318 (1993-09-01)
Monomethoxypolyethylene glycol (mPEG) of average molecular weight 5000 was transformed in a series of synthetic steps to a new activated form of PEG, a stable thiol-protected intermediary, for reaction with cysteine residues in proteins under mild conditions to produce PEG--protein
Poly (ethylene glycol) in drug delivery: pros and cons as well as potential alternatives.
Knop K, et al.
Angewandte Chemie (International Edition in English), 49(36), 6288-6308 (2010)
I L Konorova et al.
Patologicheskaia fiziologiia i eksperimental'naia terapiia, (4)(4), 7-9 (1991-07-01)
The search for antiaggregatory compounds is undertaken, as a rule, under in vitro conditions which do not reflect the dynamics of the real process. The present work deals with study of the peculiarities of the development of the collagen induced
P I Polimeni et al.
Journal of cardiovascular pharmacology, 14(3), 374-380 (1989-09-01)
The acute hemodynamic effects of an intravenously (i.v.) injected poly(ethylene oxide), Polyox WSR N-60K (dose 50 mg/kg), were studied in the open-chest rat anesthetized with sodium pentobarbital. The injectate is one of four drag-reducing polymers known to augment in vitro

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service