Skip to Content
Merck
All Photos(1)

Key Documents

PHR1957

Supelco

Tenofovir disoproxil fumarate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Tenofovir disoproxil fumarate, Bis(POC)PMPA fumarate, Bis{[(Isopropoxycarbonyl)oxy]methyl} ({[(2R)-1-(6-amino-9H-purin-9-yl)-2-propanyl]oxy}methyl)phosphonate fumarate

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$277.00

$277.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$277.00

About This Item

Empirical Formula (Hill Notation):
C19H30N5O10P · C4H4O4
CAS Number:
Molecular Weight:
635.51
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$277.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1643656

API family

tenofovir

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

OC(/C=C/C(O)=O)=O.NC1=C2C(N(C[C@@H](C)OCP(OCOC(OC(C)C)=O)(OCOC(OC(C)C)=O)=O)C=N2)=NC=N1

InChI

1S/C19H30N5O10P.C4H4O4/c1-12(2)33-18(25)28-9-31-35(27,32-10-29-19(26)34-13(3)4)11-30-14(5)6-24-8-23-15-16(20)21-7-22-17(15)24;5-3(6)1-2-4(7)8/h7-8,12-14H,6,9-11H2,1-5H3,(H2,20,21,22);1-2H,(H,5,6)(H,7,8)/b;2-1+/t14-;/m1./s1

InChI key

VCMJCVGFSROFHV-WZGZYPNHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Tenofovir disoproxil fumarate is a prodrug of tenofovir, a nucleotide analogue reverse transcriptase inhibitor (nRTI) that causes premature termination of DNA transcription. In a T cell line and primary blood lymphocytes, the antiviral activity of tenofovir disoproxil was shown to be more than 100-fold greater than tenofovir because of its rapid intracellular uptake. Tenofovir disoproxil fumarate has been used alone and in various combinations for the prevention and treatment of HIV/AIDS and chronic hepatitis B infections and is on the World Health Organization′s List of Essential Medicines.
Tenofovir disoproxil fumarate is a prodrug of tenofovir, a nucleotide analogue reverse transcriptase inhibitor (nRTI); anti-retroviral.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB2763 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - STOT RE 2

Target Organs

Bone,Kidney

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Please, what is the solubility of this drug? And what solvent?

    1 answer
    1. Product PHR1957, which is a reference material, will generally be soluble in polar solvents (methanol, chloroform, DMSO), but solubility is not a tested parameter for this product. It may also be minimally soluble in water, up to 3 mg/mL, but may require heat to go into solution.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service