Skip to Content
Merck
All Photos(1)

Key Documents

PHR1272

Supelco

Maleic acid

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

cis-Butenedioic acid, Toxilic acid

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$177.00

$177.00


Available to ship onApril 01, 2025Details


Request a Bulk Order

Select a Size

Change View
500 MG
$177.00

About This Item

Linear Formula:
HO2CCH=CHCO2H
CAS Number:
Molecular Weight:
116.07
Beilstein:
605762
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$177.00


Available to ship onApril 01, 2025Details


Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. M01000000
traceable to USP 1374500

API family

maleic acid

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

130-135 °C (lit.)

density

1.59 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

OC(=O)\C=C/C(O)=O

InChI

1S/C4H4O4/c5-3(6)1-2-4(7)8/h1-2H,(H,5,6)(H,7,8)/b2-1-

InChI key

VZCYOOQTPOCHFL-UPHRSURJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Maleic acid may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by a spectrophotometry technique.[1]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA0008 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spectrophotometric determination of ascorbic acid in pharmaceuticals by background correction and flow injection
Verma KK, et al.
Analyst, 116(6), 641-645 (1991)
Ela Lutwak et al.
Reproduction (Cambridge, England), 148(5), 507-517 (2014-08-15)
Similar expression to FGF (Sef or IL17-RD), is a tumor suppressor and an inhibitor of growth factors as well as of pro-inflammatory cytokine signaling. In this study, we examined the regulation of Sef expression by gonadotropins during ovarian folliculogenesis. In
Jianbin Zhang et al.
AAPS PharmSciTech, 15(3), 731-740 (2014-02-21)
The objective of this study is to investigate the effect of lipolysis on the release of poorly water-soluble drug from SMEDDS in the perspective of drug core/shell location. For this purpose, four SMEDDS formulations with various core/shell properties were developed
Yoonsung Lee et al.
Nature communications, 5, 5583-5583 (2014-11-28)
Haematopoietic stem cells (HSCs) derive from haemogenic endothelial cells of the primitive dorsal aorta (DA) during vertebrate embryogenesis. The molecular mechanisms governing this unique endothelial to haematopoietic transition remain unclear. Here, we demonstrate a novel requirement for fibroblast growth factor
Arul Balaji Velu et al.
Antiviral research, 112, 18-25 (2014-12-03)
Enterovirus 71 (EV71) infections can cause hand, foot, and mouth disease with severe neurological complications. Because no clinical drug is available for treating EV71 infections, developing an efficient antiviral medication against EV71 infection is crucial. This study indicated that 6-bromo-2-[1-(2,5-dimethylphenyl)-5-methyl-1H-pyrazol-4-yl]

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service