Skip to Content
Merck
All Photos(1)

Key Documents

84840

Sigma-Aldrich

Dibutyl sebacate

≥97.0% (GC)

Synonym(s):

Decanedioic acid dibutyl ester, Sebacic acid dibutyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
$110.00
1 L
$149.00

$110.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
$110.00
1 L
$149.00

About This Item

Linear Formula:
[-(CH2)4CO2(CH2)3CH3]2
CAS Number:
Molecular Weight:
314.46
Beilstein:
1798308
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$110.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.441 (lit.)
n20/D 1.441

bp

178-179 °C/3 mmHg (lit.)

density

0.936 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCCCOC(=O)CCCCCCCCC(=O)OCCCC

InChI

1S/C18H34O4/c1-3-5-15-21-17(19)13-11-9-7-8-10-12-14-18(20)22-16-6-4-2/h3-16H2,1-2H3

InChI key

PYGXAGIECVVIOZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Dibutyl sebacate has been used as a plasticizer to prepare free ethylcellulose films.[1][2] It can be utilized as a core in liquid-core capsules with cross-linked alginate/polyacrylamide membrane prepared via co-extrusion jet-break-up method for the extraction of the pesticide, atrazine.[2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

366.8 °F - open cup

Flash Point(C)

186 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Micro-encapsulated organic phase for enhanced bioremediation of hydrophobic organic pollutants.
Wyss A, et al.
Enzyme and Microbial Technology, 40(1), 25-31 (2006)
Physico-mechanical analysis of free ethylcellulose films plasticized with incremental weight percents of dibutyl sebacate.
Kangarlou S and Haririan I
Iranian Journal of Pharmaceutical Sciences, 3(3), 135-142 (2007)
T Quinten et al.
Drug development and industrial pharmacy, 37(2), 149-159 (2010-07-10)
It was the aim of the present study to develop sustained-release matrix tablets by means of injection molding of ethylcellulose (EC) and polyethylene oxide (PEO) mixtures and to evaluate the influence of process temperature, matrix composition, and viscosity grade of
Kangarlou Sogol et al.
International journal of pharmaceutics, 408(1-2), 1-8 (2010-11-26)
The object of this study was to investigate the influence of static and dynamic forces on mechanical properties of the biocompatible polymer ethyl cellulose. Similar polymeric films containing 40% (w/w) of the plasticizer dibutyl sebacate were subjected to tensile forces
Thomas Quinten et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 71(1), 145-154 (2008-05-31)
The aim of this study was to develop sustained-release matrix tablets by means of injection moulding and to evaluate the influence of process temperature, matrix composition (EC and HPMC concentration) and viscosity grade of ethylcellulose (EC) and hydroxypropylmethylcellulose (HPMC) on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service