Skip to Content
Merck
All Photos(1)

Key Documents

PHR1702

Supelco

Fluticasone Propionate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Fluticasone propionate, (6α,11β,16α,17α)-6,9-Difluoro-11-hydroxy-16-methyl-3-oxo-17-(1-oxopropoxy)androsta-1,4-diene-17-carbothioic acid S-(fluoromethyl) ester

Sign Into View Organizational & Contract Pricing

Select a Size

200 MG
$345.00

$345.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 MG
$345.00

About This Item

Empirical Formula (Hill Notation):
C25H31F3O5S
CAS Number:
Molecular Weight:
500.57
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$345.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 587
traceable to Ph. Eur. Y0000284
traceable to USP 1285873

API family

fluticasone

CofA

current certificate can be downloaded

packaging

pkg of 200 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCC(=O)O[C@@]1([C@H](C)C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)SCF

InChI

1S/C25H31F3O5S/c1-5-20(31)33-25(21(32)34-12-26)13(2)8-15-16-10-18(27)17-9-14(29)6-7-22(17,3)24(16,28)19(30)11-23(15,25)4/h6-7,9,13,15-16,18-19,30H,5,8,10-12H2,1-4H3/t13-,15+,16+,18+,19+,22+,23+,24+,25+/m1/s1

InChI key

WMWTYOKRWGGJOA-CENSZEJFSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Fluticasone Propionate, a neutral, highly potent trifluorinated corticosteroid, is a component of aqueous nasal sprays. It shows potent topical anti-inflammatory activity and is highly efficient in suppressing the nasal symptoms associated with rhinitis and asthma.[1][2]

Application

Fluticasone Propionate may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations and plasma samples[1] by various chromatography techniques.[2][3][4]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Fluticasone propionate is a second generation glucocorticoid. Used as an anti-inflammatory agent for asthma. Shown to enhance eosinophil apoptosis in a concentration-dependent manner via the glucocorticoid receptor.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB9941 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - STOT RE 2

Target Organs

Endocrine system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HPTLC-densitometric method for simultaneous determination of salmeterol xinafoate and fluticasone propionate in dry powder inhalers.
Kasaye L, et al.
Saudi Pharmaceutical Journal : SPJ : The Official Publication of the Saudi Pharmaceutical Society, 18(3), 153-159 (2003)
Validation of a stability indicating reversed phase LC method for the determination of fluticasone propionate in pharmaceutical formulations.
Sangoi MDS, et al.
Journal of Liquid Chromatography and Related Technologies, 31(14), 2113-2127 (2008)
A rapid and sensitive HPLC?APCI-MS/MS method determination of fluticasone in human plasma: Application for a bioequivalency study in nasal spray formulations.
Byrro RMD, et al.
Journal of Pharmaceutical and Biomedical Analysis, 61(5), 38-43 (2012)
Oil-in-water microemulsion high performance liquid chromatographic analysis of pharmaceuticals.
Marsh A, et al.
Chromatographia, 59(9), 531-542 (2004)
Nunzio Vicario et al.
International journal of molecular sciences, 20(8) (2019-04-26)
Sonic hedgehog (Shh) signaling is a key pathway within the central nervous system (CNS), during both development and adulthood, and its activation via the 7-transmembrane protein Smoothened (Smo) may promote neuroprotection and restoration during neurodegenerative disorders. Shh signaling may also

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service