Skip to Content
Merck
All Photos(1)

Key Documents

PHR1422

Supelco

Atorvastatin Calcium

Atorvastatin Calcium

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Atorvastatin hemicalcium salt sesquihydrate, 1H-Pyrrole-1-heptanoic acid, [R-(R*,R*)]-2-(4-flurophenyl)-β,δ-dihydroxy-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl], calcium salt (2:1)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$121.00

$121.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 15, 2025Details


Request a Bulk Order

Select a Size

Change View
1 G
$121.00

About This Item

Empirical Formula (Hill Notation):
C66H68CaF2N4O10 · 3H2O
CAS Number:
Molecular Weight:
1209.39
UNSPSC Code:
41116107
NACRES:
NA.24

$121.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 15, 2025Details


Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0001327
traceable to USP 1044516

API family

atorvastatin

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

[Ca+2].Fc5ccc(cc5)c6[n](c(c(c6c8ccccc8)C(=O)Nc7ccccc7)C(C)C)CC[C@@H](O)C[C@@H](O)CC(=O)O.Fc1ccc(cc1)c2[n](c(c(c2c4ccccc4)\C(=N\c3ccccc3)\[O-])C(C)C)CC[C@@H](O)C[C@@H](O)CC(=O)[O-].O.O.O

InChI

1S/2C33H35FN2O5.Ca.3H2O/c2*1-21(2)31-30(33(41)35-25-11-7-4-8-12-25)29(22-9-5-3-6-10-22)32(23-13-15-24(34)16-14-23)36(31)18-17-26(37)19-27(38)20-28(39)40;;;;/h2*3-16,21,26-27,37-38H,17-20H2,1-2H3,(H,35,41)(H,39,40);;3*1H2/q;;+2;;;/p-2/t2*26-,27-;;;;/m11..../s1

InChI key

SHZPNDRIDUBNMH-NIJVSVLQSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Atorvastatin Calcium is a synthetic cholesterol lowering agent,[1] which acts as an inhibitor of 3-hydroxy-3-methyl glutaryl coenzyme A reductase. It is widely used in combination with amlodipine besylate as a medication in the treatment of vasospastic angina, hypertension, chronic stable angina and primary dysbetalipoproteinemia.[2]
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Azathioprine may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations[2][1] using reversed-phase high-performance liquid chromatography (RP-HPLC)[2] and spectrophotometric techniques.[1]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC0148 in the slot below. This is an example certificate only and may not be the lot that you receive.

Analyte

Description

    Atorvastatin hemicalcium salt sesquihydrate

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Risheng Yang et al.
Oncogene, 39(40), 6340-6353 (2020-08-29)
Isocitrate dehydrogenase (IDH) mutation is the most important initiating event in gliomagenesis, and the increasing evidence shows that IDH mutation is associated with the metabolic reprogramming in the tumor. Dysregulated cholesterol metabolism is a hallmark of tumor cells, but the
Yanli Wang et al.
Histology and histopathology, 29(12), 1593-1600 (2014-08-01)
Statins are often prescribed for treatment of cardiovascular diseases, although there are still many patients who cannot be effectively treated by statins alone. Both probucol and cilostazol exhibit anti-atherogenic effects. In the current study, we attempted to investigate whether a
Simultaneous Spectrophotometric Determination of Atorvastatin Calcium and Ezetimibe in Tablet Dosage Form
GRB, et al.
International Journal of ChemTech Research, 1(2), 233-236 (2009)
Stability indicating RP-HPLC estimation of atorvastatin calcium and amlodipine besylate in pharmaceutical formulations
Shah AD, et al.
Indian Journal of Pharmaceutical Sciences, 70(6), 754-754 (2008)
Xiao-ling Li et al.
International immunopharmacology, 23(2), 546-549 (2014-12-03)
This study aimed to investigate the efficacy of combined atorvastatin calcium and methylprednisolone for the treatment of multiple sclerosis relapse. Patients with multiple sclerosis (MS) at the relapse phase were randomized to receive either combined treatment of atorvastatin calcium and

Questions

1–4 of 4 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. I have made a stock solitution in DMSO. At what temperature should I store it and what is the shelf life?

    1 answer
    1. The stability of this product in DMSO has not been determined. However, various sources recommend storing stock solutions in DMSO at -20°C for 1 month and at -80°C for 1 year. As a general rule, avoid repeated freeze/thaw cycles. This information has not been validated.

      Helpful?

  4. Buenos días, como debe reconstituirse el producto: Atorvastatin Calcium, PHR1422? Muchas gracias, Aina

    1 answer
    1. The following solubility characteristics are given in the monograph of the European Pharmacopoeia: practically insoluble in water, very slightly soluble to very soluble in ethanol (96 percent), freely soluble to very soluble in methanol, practically insoluble to freely soluble in methylene chloride, and freely soluble in dimethyl sulfoxide.

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service