Skip to Content
Merck
All Photos(1)

Key Documents

PHR1619

Supelco

Beclomethasone Dipropionate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Beclomethasone dipropionate, 9-Chloro-11β,17,21-trihydroxy-16β-methylpregna-1,4-diene-3,20-dione 17,21-dipropionate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$225.00

$225.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 14, 2025Details


Request a Bulk Order

Select a Size

Change View
1 G
$225.00

About This Item

Empirical Formula (Hill Notation):
C28H37ClO7
CAS Number:
Molecular Weight:
521.04
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$225.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 14, 2025Details


Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 30
traceable to Ph. Eur. B0305000
traceable to USP 1048506

API family

beclometasone

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CCC(=O)OCC(=O)[C@@]1(OC(=O)CC)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]12C

InChI

1S/C28H37ClO7/c1-6-23(33)35-15-22(32)28(36-24(34)7-2)16(3)12-20-19-9-8-17-13-18(30)10-11-25(17,4)27(19,29)21(31)14-26(20,28)5/h10-11,13,16,19-21,31H,6-9,12,14-15H2,1-5H3/t16-,19-,20-,21-,25-,26-,27-,28-/m0/s1

InChI key

KUVIULQEHSCUHY-XYWKZLDCSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

Beclomethasone Dipropionate is a second generation[1] anti-inflammatory corticosteroid used for the treatment of allergic rhinitis.[2]
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Beclomethasone Dipropionate may be used as a pharmaceutical secondary reference standard for the determination of the analyte in biological fluids[3] and ear drops formulations[4] by chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC2965 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - STOT RE 2

Target Organs

Adrenal gland,Immune system,Bone

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of beclomethasone, beclomethasone monopropionate and beclomethasone dipropionate in biological fluids using a particle beam interface for combining liquid chromatography with negative-ion chemical ionization mass spectrometry
Girault J, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 564(1), 43-53 (1991)
Efficacy and safety of beclomethasone dipropionate nasal aerosol in children with perennial allergic rhinitis
Berger WE, et al.
Annals of Allergy, Asthma & Immunology, 115(2), 130-136 (2015)
Efficacy and safety of oral beclomethasone dipropionate in ulcerative colitis: a systematic review and meta-analysis
Manguso F, et al.
Testing, 11(11), e0166455-e0166455 (2016)
Determination of beclomethasone dipropionate, clotrimazole, chloramphenicol and lidocaine in pharmaceutical formulations using a novel RP-HPLC method
Reddy MU, et al.
International Journal of Pharma and Bio Sciences, 2(1), 453-462 (2011)
Determination of beclomethasone dipropionate, clotrimazole, chloramphenicol and lidocaine in pharmaceutical formulations using a novel RP-HPLC method
Reddy MU, et al.
International Journal of Pharmacy and Biological Sciences, 2(1), 453-462 (2011)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service