Skip to Content
Merck
All Photos(1)

Documents

PHR1655

Supelco

Carbidopa

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

(S)-3-(3,4-Dihydroxyphenyl)-2-hydrazino-2-methylpropionic acid monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2O4 · H2O
CAS Number:
Molecular Weight:
244.24
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 60
traceable to Ph. Eur. C0460000
traceable to USP 1095506

API family

carbidopa

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

O.C[C@@](Cc1ccc(O)c(O)c1)(NN)C(O)=O

InChI

1S/C10H14N2O4.H2O/c1-10(12-11,9(15)16)5-6-2-3-7(13)8(14)4-6;/h2-4,12-14H,5,11H2,1H3,(H,15,16);1H2/t10-;/m0./s1

InChI key

QTAOMKOIBXZKND-PPHPATTJSA-N

Gene Information

human ... DDC(1644)

Looking for similar products? Visit Product Comparison Guide

General description

Carbidopa is a peripheral decarboxylase inhibitor which is often formulated with levodopa for the management of the symptoms of Parkinson′s disease. Its mode of action involves the reduction of systemic production of dopamine and minimization of undesirable effects such as nausea.
Pharmaceutical secondary standard for application in quality control. Provides pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Carbidopa may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA5756 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chaotropic agents in liquid chromatographic method development for the simultaneous analysis of levodopa, carbidopa, entacapone and their impurities
Vemic A, et al.
Journal of Pharmaceutical and Biomedical Analysis, 77(1-2), 9-15 (2013)
Stability-indicating method for the determination of levodopa, levodopa-carbidopa and related impurities
Kafil JB and Dhingra BS
Journal of Chromatography A, 667(1-2), 175-181 (1994)
Modified high-performance liquid chromatography with electrochemical detection method for plasma measurement of levodopa, 3-O-methyldopa, dopamine, carbidopa and 3, 4-dihydroxyphenyl acetic acid
Karimi M, et al.
Journal of Chromatography. B, Biomedical Applications, 836(1-2), 120-123 (2006)
Simultaneous estimation of levodopa and carbidopa by RP-HPLC using a fluorescence detector: its application to a pharmaceutical dosage form
Raut PP and Charde SY
Luminescence, 29(7), 762-771 (2014)
Vayu Maini Rekdal et al.
Science (New York, N.Y.), 364(6445) (2019-06-15)
The human gut microbiota metabolizes the Parkinson's disease medication Levodopa (l-dopa), potentially reducing drug availability and causing side effects. However, the organisms, genes, and enzymes responsible for this activity in patients and their susceptibility to inhibition by host-targeted drugs are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service