Skip to Content
Merck
All Photos(1)

Key Documents

796883

Sigma-Aldrich

3,4-dihydroxyphenylacetone

95%

Synonym(s):

1-(3,4-Dihydroxyphenyl)-2-propanone, 1-(3′,4′-Dihydroxyphenyl)-2-propanone

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$161.00

$161.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$161.00

About This Item

Empirical Formula (Hill Notation):
C9H10O3
CAS Number:
Molecular Weight:
166.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$161.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

95%

form

powder

functional group

ketone

SMILES string

CC(CC1=CC(O)=C(O)C=C1)=O

InChI

1S/C9H10O3/c1-6(10)4-7-2-3-8(11)9(12)5-7/h2-3,5,11-12H,4H2,1H3

InChI key

JQXBETDGCMQLMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,4-dihydroxyphenylacetoneis an alkylbenzene. 3,4-dihydroxyphenylacetone is the product of oxidative deamination of α-methylDopa. The product is intended for forensic and research purposes.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mammalian Dopa decarboxylase: structure, catalytic activity and inhibition
Bertoldi M.
Archives of Biochemistry and Biophysics, 546, 1-7 (2014)
Mohammadreza Shariatgorji et al.
Nature methods, 16(10), 1021-1028 (2019-09-25)
We present a mass spectrometry imaging (MSI) approach for the comprehensive mapping of neurotransmitter networks in specific brain regions. Our fluoromethylpyridinium-based reactive matrices facilitate the covalent charge-tagging of molecules containing phenolic hydroxyl and/or primary or secondary amine groups, including dopaminergic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service