Skip to Content
Merck
All Photos(1)

Key Documents

PHR1090

Supelco

Dopamine hydrochloride

Dopamine hydrochloride

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

2-(3,4-Dihydroxyphenyl)ethylamine hydrochloride, 3,4-Dihydroxyphenethylamine hydrochloride, 3-Hydroxytyramine hydrochloride, 4-(2-Aminoethyl)-1,2-benzenediol hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$126.00

$126.00


Estimated to ship onMarch 19, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$126.00

About This Item

Linear Formula:
(HO)2C6H3CH2CH2NH2·HCl
CAS Number:
Molecular Weight:
189.64
Beilstein:
3656720
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$126.00


Estimated to ship onMarch 19, 2025


Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 468
traceable to Ph. Eur. D2690000
traceable to USP 1225204

API family

dopamine

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

248-250 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Cl[H].NCCc1ccc(O)c(O)c1

InChI

1S/C8H11NO2.ClH/c9-4-3-6-1-2-7(10)8(11)5-6;/h1-2,5,10-11H,3-4,9H2;1H

InChI key

CTENFNNZBMHDDG-UHFFFAOYSA-N

Gene Information

human ... ADRB1(153)

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Dopamine is a neurotransmitter which is a naturally occurring catecholamine. Dopamine hydrochloride salt is indicated as a medicine for the treatment of acute congestive and renal failures.[1]

Application

Dopamine hydrochloride may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by spectrophotometry,[1][2] colorimetry and chromatography.[3]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA7131 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Intropin (dopamine hydrochloride) intravenous admixture compatibility
Gardella LA, et al.
American Journal of Hospital Pharmacy, 32(6), 575-578 (1975)
Rapid Spectrophotometric Determination Of Dopamine Hydrochloride With Chloramine - T
Nagaraja P, et al.
Indian Journal of Pharmaceutical Sciences, 60(2), 99-99 (2016)
Spectrophotometric methods for the determination of certain catecholamine derivatives in pharmaceutical preparations
Nagaraja P, et al.
Talanta, 46(1), 39-44 (1998)
Darae Kang et al.
Poultry science, 100(3), 100964-100964 (2021-03-04)
The effects of early heat conditioning on the acute heat stress response in broilers were investigated via the growth performance, dopamine, serotonin, and corticosterone and the expression of heat shock proteins (HSP) and heat shock factors. One-day-old chicks (n =
A Jon Stoessl et al.
Lancet (London, England), 384(9942), 532-544 (2014-06-24)
Recent advances in structural and functional imaging have greatly improved our ability to assess normal functions of the basal ganglia, diagnose parkinsonian syndromes, understand the pathophysiology of parkinsonism and other movement disorders, and detect and monitor disease progression. Radionuclide imaging

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service