Skip to Content
Merck
Dopamine hydrochloride certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Dopamine hydrochloride

Dopamine hydrochloride certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
2-(3,4-Dihydroxyphenyl)ethylamine hydrochloride, 3,4-Dihydroxyphenethylamine hydrochloride, 3-Hydroxytyramine hydrochloride, 4-(2-Aminoethyl)-1,2-benzenediol hydrochloride
Linear Formula:
(HO)2C6H3CH2CH2NH2·HCl
CAS Number:
Molecular Weight:
189.64
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Agency
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique