Skip to Content
Merck
All Photos(1)

Key Documents

79214

Sigma-Aldrich

Bacteria stain propidium iodide solution

1 mg/mL in H2O, ready to use solution, 100 assays

Synonym(s):

Propidium iodide solution, 3,8-Diamino-5-(3-diethylaminopropyl)-6-phenylphenanthridinium iodide methiodide, 3,8-Diamino-5-[3-(diethylmethylammonio)propyl]-6-phenylphenanthridinium, diiodide

Sign Into View Organizational & Contract Pricing

Select a Size

4 X 25 μL
$349.00

$349.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 30, 2025


Request a Bulk Order

Select a Size

Change View
4 X 25 μL
$349.00

About This Item

Empirical Formula (Hill Notation):
C27H34I2N4
CAS Number:
Molecular Weight:
668.39
Beilstein:
3843838
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.32

$349.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 30, 2025


Request a Bulk Order

Assay

≥90.0% (HPLC)

form

liquid

quality

ready to use solution, 100 assays

concentration

1 mg/mL in H2O

fluorescence

λex 530 nm; λem 620 nm±5 nm in 0.1 M phosphate pH 7.0

storage temp.

−20°C

SMILES string

[I-].[I-].CC[N+](C)(CC)CCC[n+]1c(-c2ccccc2)c3cc(N)ccc3c4ccc(N)cc14

InChI

1S/C27H33N4.2HI/c1-4-31(3,5-2)17-9-16-30-26-19-22(29)13-15-24(26)23-14-12-21(28)18-25(23)27(30)20-10-7-6-8-11-20;;/h6-8,10-15,18-19,29H,4-5,9,16-17,28H2,1-3H3;2*1H/q+1;;/p-1

InChI key

XJMOSONTPMZWPB-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Membrane impermeant, phenanthridinium intercalator dye excited by mercury or xenon-arc lamps or argon-ion laser. Suitable for use in fluorescence microscopy, confocal laser scanning microscopy, flow cytometry, and fluorometry. Used as a nuclear counterstain.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service