Skip to Content
Merck
All Photos(3)

Documents

06432379001

Roche

Cell Viability Imaging Kit 5x96

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.55

usage

sufficient for 5 96-well plate(s)

Quality Level

packaging

pkg of 1 kit

manufacturer/tradename

Roche

storage condition

protect from light

fluorescence

λex 361 nm; λem 486 nm (nuclei dye)
λex 490 nm; λem 515 nm (viable dye)
λex 535 nm; λem 617 nm (dead dye)

detection method

fluorometric

shipped in

dry ice

storage temp.

−20°C (−15°C to −25°C)

General description

The Cell Viability Imaging Kit is a simple and rapid way to measure mammalian cell viability using fluorescence microscopy and automated imaging platforms, such as the Cellavista System. Optimized for 96-well plates, the numbers of viable and dead cells, as well as total cell numbers, are determined simultaneously for the area of the well selected.
The kit is designed for 5 × 96 reactions for use in medium to high throughput analysis of cell behavior.
  • Determine total cell number using fluorescence staining of cell nuclei.
  • Fluorimetrically determine the percentage of dead cells based on membrane permeability.
  • Verify the labeling of viable cells quantified by fluorescence staining using a vital dye

Application

Cell quantification and cell viability control are very important methods in cell biology. This includes both life science research and industrial applications. The simple, accurate and fast “one step assay” makes the Cell Viability Imaging Kit an ideal tool for quality control of cells in cellular workflows, especially for medium to high throughput applications.

Features and Benefits

Cell counting and cell viability are essential for cellular workflows. The Roche Cell Viability Imaging Kit meets the following requirements:
  • No risk of losing cells in the one step assay: There are no fixation or washing steps.
  • Save time and resources: Total assay time for a 1 x 96 well microplate is only 35 minutes.
  • Obtain highly reproducible results using automated imaging and data evaluation with Roche′s Cellavista System.

Packaging

1 kit containing 3 components.

Preparation Note

Storage conditions (working solution): Vial 3: The product is stable until the expiration date printed on the label when stored at -15 to -25 °C. Contents can be frozen and thawed at least 5 times after preparation of solution.

Storage and Stability

Store at -15–-25 °C. (Vial 1 and Vial 2 can be frozen and thawed at least 5 times. Vial 3 can be frozen and thawed at least 5 times after preparation of solution.)

Other Notes

For life science research only. Not for use in diagnostic procedures.

Kit Components Only

Product No.
Description

  • Nuclei Dye (blue cap) for fluorimetric determination of cell count based on labeling of cell nuclei

  • Dead Dye (red cap) for fluorimetric determination of dead cells based on cell permeability

  • Viable Dye ( green cap) for fluorimetric determination of viable cells based on metabolic activity

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Muta. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fengliang Wang et al.
Molecular medicine reports, 16(4), 5441-5449 (2017-08-30)
Previous studies have reported that angelicin exerted antiproliferative effects on several types of tumor cell. However, to the best of our knowledge, the effects of angelicin monotherapy on human liver cancer remain to be investigated. In the present study, the
Lingbo Xu et al.
Molecular medicine reports, 23(6) (2021-04-22)
Our previous study reported that microRNA (miR)‑30a‑5p upregulation under hypoxia postconditioning (HPostC) exert a protective effect on aged H9C2 cells against hypoxia/reoxygenation injury via DNA methyltransferase 3B‑induced DNA hypomethylation at the miR‑30a‑5p gene promoter. This suggests that miR‑30a‑5p may be
Kun Xiao et al.
Gut pathogens, 13(1), 63-63 (2021-10-21)
The liver plays an important role in production and metabolism of homocysteine (Hcy), which has been reported to be involved in liver injury. In our previous work, we confirm that Hcy can induce liver injury by activating endoplasmic reticulum (ER)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service