Skip to Content
Merck
All Photos(2)

Key Documents

NIST2897A

Ethanol solution

NIST® SRM® 2897a, nominal mass fraction 2%

Synonym(s):

Ethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OH
CAS Number:
Molecular Weight:
46.07
Beilstein:
1718733
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

Agency

NIST standard reference material

packaging

pkg of 5 x 10 mL

manufacturer/tradename

NIST®

concentration

(nominal mass fraction 2%)

technique(s)

Karl-Fischer titration: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

SMILES string

CCO

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

One unit of SRM 2897a comprises five 10-milliliter ampoules, with each consisting of ~ 10 mL of solution.

SRM 2897A_cert

SRM 2897A_SDS

Application

This SRM is intended for calibrating the instruments and techniques used for ethanol determination.

Features and Benefits

  • Available with the certificate documenting NIST-certified mass fraction values for ethanol obtained through gravimetric solution preparation.
  • The certified values are metrologically traceable to the SI derived unit for mass fraction, expressed as percent.
  • The NIST certificate is provided with expiration certificate, storage, handling, use, and maintenance instructions.

Other Notes

Certified for the analytes listed below.
Ethanol (C2H6O)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service