Skip to Content
Merck
All Photos(1)

Key Documents

324523

Sigma-Aldrich

Ethanol-1-13C

99 atom % 13C

Synonym(s):

Ethyl alcohol-1-13C

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$1,790.00

$1,790.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
1 G
$1,790.00

About This Item

Linear Formula:
CH313CH2OH
CAS Number:
Molecular Weight:
47.06
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.12

$1,790.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

99 atom % 13C

Assay

99% (CP)

refractive index

n20/D 1.362 (lit.)

bp

78 °C (lit.)

mp

-114 °C (lit.)

density

0.806 g/mL at 25 °C

mass shift

M+1

SMILES string

C[13CH2]O

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3/i2+1

InChI key

LFQSCWFLJHTTHZ-VQEHIDDOSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul J Fitzgerald
International journal of molecular sciences, 22(19) (2021-10-14)
It is well established that a wide range of drugs of abuse acutely boost the signaling of the sympathetic nervous system and the hypothalamic-pituitary-adrenal (HPA) axis, where norepinephrine and epinephrine are major output molecules. This stimulatory effect is accompanied by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service