Skip to Content
Merck
All Photos(2)

Key Documents

NIST2898A

Ethanol solution

NIST® SRM® 2898a, nominal mass fraction 6%

Synonym(s):

Ethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OH
CAS Number:
Molecular Weight:
46.07
Beilstein:
1718733
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

packaging

pkg of 5 x 10 mL

manufacturer/tradename

NIST®

concentration

(nominal mass fraction 6%)

technique(s)

Karl-Fischer titration: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

SMILES string

CCO

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Each unit of SRM 2898a contains five 10-milliliter ampoules, with each ampoule containing ~ 10 mL of solution. For more information, please refer to the SDS and COA.

SRM 2898A_cert

SRM 2898A_SDS

Application

This Standard Reference Material (SRM) is a solution composed of ethanol (ethyl alcohol) in water, with a nominal mass fraction of 6%. It is primarily intended for the calibration of instruments and techniques used in the determination of ethanol.

Features and Benefits

  • The certified mass fraction value is derived from results obtained through the gravimetric preparation of the solution and analytical data determined via gas chromatography.
  • Maintenance and expiration of SRM certification, instructions for handling, storage, and use are provided on NIST certificate.

Other Notes

Certified for the analytes listed below.
Ethanol-Water Solution

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Hélène Alami-Durante et al.
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology, 242, 110652-110652 (2020-01-11)
Two experiments were conducted to test if manipulations of the Arginine-Nitric oxide pathway during the early life of rainbow trout would act on its early myogenic process. In experiment 1, trout embryos were immersed at 72° days post-fertilization (°dpf) or
Laurent L'homme et al.
Scientific reports, 10(1), 21095-21095 (2020-12-05)
Retinoic acid receptor-related orphan receptor-alpha (RORα) is a transcription factor from the nuclear receptor family expressed by immune cells and involved in the development of obesity, insulin resistance (IR) and non-alcoholic steatohepatitis (NASH). It was recently reported that mice deficient

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service