Skip to Content
Merck
All Photos(1)

Documents

NIST158A

Silicon bronze

NIST® SRM® 158a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

Quality Level

form

chips

packaging

pkg of 150 g

manufacturer/tradename

NIST®

General description

Other Notes

See certificate for values and more details at nist.gov/SRM.
Aluminum (Al), Copper (Cu), Iron (Fe), Lead (Pb), Manganese (Mn), Phosphorus (P), Silicon (Si), Tin (Sn), Zinc (Zn)

Reference/Informational value is provided for the following analytes.
Chromium (Cr), Nickel (Ni), Silver (Ag)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service