Skip to Content
Merck
All Photos(1)

Key Documents

412805

Sigma-Aldrich

4,4′-Methylenebis(N,N-diglycidylaniline)

Synonym(s):

4,4′-Methylenebis[N ,N -bis(oxiranylmethyl)aniline], 4,4′-Methylenedianiline tetraglycidyl ether, N ,N ,N ′,N ′-Tetraglycidyl-4,4′-methylenebisbenzenamine, N ,N ,N ′,N ′-Tetraglycidyl-4,4′-methylenedianiline, Bis[4-(diglycidylamino)phenyl]methane

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$102.00
500 ML
$355.00

$102.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$102.00
500 ML
$355.00

About This Item

Empirical Formula (Hill Notation):
C25H30N2O4
CAS Number:
Molecular Weight:
422.52
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$102.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

viscous liquid

Quality Level

refractive index

n20/D 1.601 (lit.)

density

1.15 g/mL at 25 °C (lit.)

SMILES string

C1OC1CN(CC2CO2)c3ccc(Cc4ccc(cc4)N(CC5CO5)CC6CO6)cc3

InChI

1S/C25H30N2O4/c1-5-20(26(10-22-14-28-22)11-23-15-29-23)6-2-18(1)9-19-3-7-21(8-4-19)27(12-24-16-30-24)13-25-17-31-25/h1-8,22-25H,9-17H2

InChI key

FAUAZXVRLVIARB-UHFFFAOYSA-N

General description

4,4′-Methylenebis(N,N-diglycidylaniline) (MDGA) is a bifunctional monomer possessing two epoxy groups (glycidyl groups) and two amine groups. It is commonly used as a monomer in the production of thermosetting polymers, particularly epoxy resins. MDGA-based epoxy resins can exhibit desirable properties such as high strength, chemical resistance, and thermal stability, which makes them suitable for a wide range of applications such as adhesives, coatings, composites, and electronic encapsulation.[1][2][3]

Application

4,4′-Methylenebis(N,N-diglycidylaniline) can be used:
  • As a starting material to synthesize UV-curable tetra-functional epoxy acrylate (EA4), which is used as a crosslinker for UV-curable resins.
  • In the synthesis of bismaleimide/diallyl bisphenol A (BMI/DBA)–epoxy interpenetrating network resins, which have potential applications in the aerospace and automotive industries because of their high thermal stability and low activation energy.
  • As a crosslinking agent in the production of biocompatible materials, such as hydrogels and other tissue engineering scaffolds. Its ability to form strong and stable crosslinks makes it valuable in the creation of medical devices, implants, and drug delivery systems. Additionally, it is utilized in the synthesis of biocompatible polymers and materials with tailored properties for biomedical applications.
4,4′-Methylenebis(N,N-diglycidylaniline) can be also used as a precursor to synthesize:
  • Poly(hexamethylene biguanide) based polymer networks which are applicable as catalysts for the transesterification of vegetable oils.
  • Tetra-functional epoxy-acrylate UV curable resins.
It can also be used as a crosslinker to functionalize carbon nanotubes to improve their mechanical strength.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Muta. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Burrows et al.
Contact dermatitis, 11(2), 80-82 (1984-08-01)
An outbreak of dermatitis occurred in an aircraft factory using epoxy resin composite material. Of 25 operatives, 14 gave positive patch test reactions to the composite material and/or diglycidylether of bisphenol A (DGEBA), tetraglycidyl-4,4'-methylene dianiline (TGMDA), and o-diglycidyl phthalate. This
L Kanerva et al.
Dermatology (Basel, Switzerland), 201(1), 29-33 (2000-09-06)
Very little is known about allergic contact dermatitis (ACD) from preimpregnated epoxy products (prepregs). To describe a patient with occupational ACD from prepregs, and report new quantitative data on the content of prepregs. A laminator developed work-related vesicular hand dermatitis.
Michael Giebler et al.
Polymers, 12(5) (2020-05-21)
Epoxy-anhydride vitrimers are covalent adaptable networks, which undergo associative bond exchange reactions at elevated temperature. Their service temperature is influenced by the glass transition temperature (Tg) as well as the topology freezing transition temperature (Tv), at which the covalent bond
F Ayala et al.
Contact dermatitis, 22(5), 262-266 (1990-05-01)
Thin-layer chromatography, column chromatography, mass spectrometry and nuclear magnetic resonance were used to investigate the chemical constituents and haptens responsible in 2 different circumstances where allergic contact dermatitis from composite materials was suspected. In an aircraft factory where epoxy resins

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service