Skip to Content
Merck
All Photos(1)

Key Documents

ERMAE701

Calcium 41Ca / 40Ca (certified for isotope abundance ratio)

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
$1,490.00

$1,490.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 SET
$1,490.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$1,490.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

certified reference material

Agency

ERM®

isotopic purity

 40Ca

manufacturer/tradename

JRC

format

neat

Analysis Note

For more information please see:
ERMAE701

Legal Information

ERM is a registered trademark of European Commission

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service