Skip to Content
Merck
All Photos(4)

Key Documents

D1033

Sigma-Aldrich

Monoclonal Anti-Desmin antibody produced in mouse

clone DE-U-10, ascites fluid

Synonym(s):

Anti-CDCD3, Anti-CSM1, Anti-CSM2, Anti-LGMD1D, Anti-LGMD1E, Anti-LGMD2R

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

DE-U-10, monoclonal

mol wt

antigen 50-55 kDa

contains

15 mM sodium azide

species reactivity

rabbit, chicken, hamster, feline, lizard, sheep, rat, human, bovine, viper, mouse, goat

technique(s)

indirect immunofluorescence: 1:40 using formalin-fixed, paraffin-embedded sections of human tissue
microarray: suitable
western blot: 1:200 using chicken gizzard extract/whole extract of mouse myoblast C2C12 cells

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Des(13346)
rat ... Des(64362)

General description

Desmin is the protein subunit of muscle-type intermediate filaments and is found predominantly in skeletal, cardiac, and smooth muscle.
Monoclonal Anti-Desmin (mouse IgG1 isotype) is derived from the hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse. The desmin protein contains a head domain, four α-helical segments, three non-helical rod segments and a tail domain. It exists as dimer and tetramer assemblies. Desmin is is present in the periphery of Z-disk in smooth and striated muscles.

Specificity

The antibody localizes desmin in cultured cells and tissue sections from several species and in tumors derived from muscle tissue (leiomyomas, rhabdomyosarcomas).

Immunogen

desmin from pig stomach

Application

Monoclonal Anti-Desmin antibody produced in mouse has been used in:
  • immunohistochemical staining
  • immunoblot analyses
  • immunocytochemistry
  • immunocytochemical localization of intermediate filaments

Monoclonal Anti-Desmin antibody produced in mouse is suitable for indirect immunofluorescence at a working dilution of 1:20 using formalin-fixed, paraffin-embedded sections of human tissue, microarray and western blotting. It may be used for immunocytochemical localization of intermediate filaments of the desmin group in all types of muscle cells and to localize desmin at the periphery of z-discs. It specifically stains desmin when used in immunoblotting assays. It may be used for labelling desmin in tumors derived from muscle tissue (leiomyomas and rhabdomyosarcomas). It was used for immunohistochemical staining of cardiac cells in a study to understand cardiac conduction through engineered tissue. It was used as a primary antibody for the detection of expression of desmin in cultured cells to study the effect of mutation in desmin gene.

Biochem/physiol Actions

Desmin provides mechanical strength to muscle tissues. Mutations in desmin gene contributes to imbalance in skeletal proteins, resulting in desminopathies. Mutations in this gene damages the stability of intermediary filaments in cardiomyocytes.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functional characterization of the novel DES mutation p. L136P associated with dilated cardiomyopathy reveals a dominant filament assembly defect
Brodehl A, et al.
Journal of Molecular and Cellular Cardiology, 91, 207-214 (2016)
Cardiac conduction through engineered tissue
Choi YH, et al.
The American Journal of Pathology, 169(1), 72-85 (2006)
Myogenic differentiation potential of human tonsil-derived mesenchymal stem cells and their potential for use to promote skeletal muscle regeneration
Park S, et al.
International Journal of Molecular Medicine, 37(5), 1209-1220 (2016)
Phenotypic expression of a novel desmin gene mutation: hypertrophic cardiomyopathy followed by systemic myopathy.
Harada H, et al.
Journal of Human Genetics, 63(2), 249-249 (2018)
Desmin: a major intermediate filament protein essential for the structural integrity and function of muscle.
Paulin D and Li Z
Experimental Cell Research, 301(1), 1-7 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service