Skip to Content
Merck
All Photos(4)

Key Documents

C8356

Sigma-Aldrich

Calcium lactate pentahydrate

meets USP testing specifications

Synonym(s):

Calcium L-lactate pentahydrate, L-Lactic acid calcium salt

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$290.00
500 G
$563.00
1 KG
$861.00

$290.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 07, 2025Details


Request a Bulk Order

Select a Size

Change View
250 G
$290.00
500 G
$563.00
1 KG
$861.00

About This Item

Linear Formula:
[CH3CH(OH)COO]2Ca · 5H2O
CAS Number:
Molecular Weight:
308.29
Beilstein:
4923285
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.21

$290.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 07, 2025Details


Request a Bulk Order

Agency

meets USP testing specifications

Quality Level

form

solid

application(s)

pharmaceutical (small molecule)

storage temp.

2-8°C

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].C[C@H](O)C(=O)O[Ca]OC(=O)[C@H](C)O

InChI

1S/2C3H6O3.Ca.5H2O/c2*1-2(4)3(5)6;;;;;;/h2*2,4H,1H3,(H,5,6);;5*1H2/q;;+2;;;;;/p-2/t2*2-;;;;;;/m00....../s1

InChI key

HPVJXNNKHRNBOY-VFGZHDCSSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Biodegradable Polymer Synthesis: Research into biodegradable polymer synthesis using Calcium Lactate Pentahydrate has shown promising results in developing eco-friendly materials with applications in medical implants and drug delivery systems. The study focuses on its role in enhancing polymer cross-linking and biocompatibility (Szterner et al., 2022).

  • Calcium Lactate Pentahydrate Safety and Solubility in Pharmaceutical Formulations: Research confirms the safety profile of Calcium Lactate Pentahydrate, emphasizing its high solubility which makes it an excellent candidate for pharmaceutical applications, particularly in mineral supplements and antacid formulations (Chaerun et al., 2020).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mostafa Seifan et al.
Applied microbiology and biotechnology, 100(23), 9895-9906 (2016-07-10)
Microbially induced calcium carbonate precipitation is an emerging process for the production of self-healing concrete. This study was aimed to investigate the effects and optimum conditions on calcium carbonate biosynthesis. Bacillus licheniformis, Bacillus sphaericus, yeast extract, urea, calcium chloride and
Taeyeon Kim et al.
Applied microbiology and biotechnology, 101(7), 2821-2830 (2017-01-13)
Conversion of crude glycerol derived from biodiesel processes to value-added chemicals has attracted much attention. Herein, Raoultella ornithinolytica B6 was investigated for the high production of 2,3-butanediol (2,3-BD) from glycerol without 1,3-propanediol (1,3-PD) formation, a by-product hindering 2,3-BD purification. By
Theresa Maier et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 131, 119-126 (2019-02-03)
To aid oral therapeutic administration to infants, a novel delivery technology, referred to as a Therapeutic Nipple Shield (TNS), was previously developed. It consists of a silicone nipple shield device and a dosage form containing a therapeutic (or Active Pharmaceutical
Qianru Hui et al.
Journal of the science of food and agriculture, 98(2), 644-651 (2017-07-01)
Phytic acid as a phosphorus storage vault provides phosphorus for plant development. It is an anti-nutritional factor for humans and some animals. However, its degradation products lower inositol phosphates have positive effects on human health. In this study, the effect
Ionuț Ledeți et al.
Pharmaceutics, 12(1) (2020-01-16)
The influence of excipients on the stability of sodium levothyroxine pentahydrate (LTSS) under ambient conditions and thermal stress was evaluated. Since LTSS is a synthetic hormone with a narrow therapeutic index, the interactions of LTSS with excipients can lead to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service