Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Calcium lactate pentahydrate
Synonym(s):
Calcium lactate pentahydrate, Calcium L-lactate pentahydrate, L-Lactic acid calcium salt
Linear Formula:
[CH
3
CH(OH)COO]
2
Ca · 5H
2
O
CAS Number:
5743-47-5
Molecular Weight:
308.29
EC Number:
248-953-3
Products
Product Category
salts (1)
Brand
SAFC (1)
Sigma-Aldrich (1)
Feature
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Application
Optical Isomer
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
1.02102
Calcium lactate pentahydrate,
EMPROVE
®
ESSENTIAL Ph Eur,BP,USP,E 327
Expand
View Pricing
C8356
Calcium lactate pentahydrate,
meets USP testing specifications
Expand
View Pricing