Skip to Content
Merck
All Photos(1)

Key Documents

21175

Sigma-Aldrich

Calcium L-lactate pentahydrate

tested according to Ph. Eur.

Synonym(s):

Calcii lactas pentahydricus, L-Lactic acid calcium salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
$382.00

$382.00


Estimated to ship onApril 15, 2025


Request a Bulk Order

Select a Size

Change View
1 KG
$382.00

About This Item

Linear Formula:
[CH3CH(OH)COO]2Ca · 5H2O
CAS Number:
Molecular Weight:
308.29
Beilstein:
4923285
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.21

$382.00


Estimated to ship onApril 15, 2025


Request a Bulk Order

Agency

USP/NF
tested according to Ph. Eur.

Quality Level

form

solid

application(s)

pharmaceutical (small molecule)

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].C[C@H](O)C(=O)O[Ca]OC(=O)[C@H](C)O

InChI

1S/2C3H6O3.Ca.5H2O/c2*1-2(4)3(5)6;;;;;;/h2*2,4H,1H3,(H,5,6);;5*1H2/q;;+2;;;;;/p-2/t2*2-;;;;;;/m00....../s1

InChI key

HPVJXNNKHRNBOY-VFGZHDCSSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Calcium L-lactate pentahydrate has been used for in vitro maturation (IVM) of horse oocytes.[1] It has also been used as a supplement for medium TCM-199.[2]

Biochem/physiol Actions

Calcium L-lactate also referred to as L-Lactic acid calcium salt or calcii lactas pentahydricus enhances the calcium bioavailability.[3] It is useful in treating calcium deficiencies.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martina Vavrusova et al.
Food & function, 5(1), 85-91 (2013-11-28)
Continuing dissolution of solid calcium L-lactate pentahydrate in saturated aqueous solutions following addition of solid sodium D-gluconate corresponding to a gluconate/lactate ratio around three was found to result in homogeneous solutions supersaturated with calcium D-gluconate by a factor of seven
Barbara Ambruosi et al.
PloS one, 6(11), e27452-e27452 (2011-11-15)
Phthalates are ubiquitous environmental contaminants because of their use in plastics and other common consumer products. Di-(2-ethylhexyl) phthalate (DEHP) is the most abundant phthalate and it impairs fertility by acting as an endocrine disruptor. The aim of the present study
Anna Lange Consiglio et al.
Reproductive biology and endocrinology : RB&E, 7, 113-113 (2009-10-20)
The identification of the adipocyte-derived obesity gene product, leptin (Ob), and subsequently its association with reproduction in rodents and humans led to speculations that leptin may be involved in the regulation of oocyte and preimplantation embryo development. In mice and
Ehsan Rezabeigi et al.
Materials science & engineering. C, Materials for biological applications, 40, 248-252 (2014-05-27)
More than four decades after the discovery of 45S5 Bioglass® as the first bioactive material, this composition is still one of the most promising materials in the tissue engineering field. Sol-gel-derived bioactive glasses generally possess improved properties over other bioactive

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service