Skip to Content
Merck
All Photos(1)

Key Documents

BCR539

Plastic film (overall migration in olive oil, film C)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$362.00

$362.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

Select a Size

Change View
1 EA
$362.00

About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

$362.00


Estimated to ship onMay 26, 2025


Request a Bulk Order

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

application(s)

chemicals and industrial polymers

format

matrix material

storage temp.

2-8°C

Application

This CRM consists of additive free linear low density polyethylene sheets. It is intended to determine the overall migration applying the draft ENV 1186 standard procedure established by the Technical Committee 194 of the European Committee for Standardisation (CEN).

Overall migration by pouch in olive oil 10 days at 40°C : 6.1 ± 1.0 (mg/dm2)

Packaging

PET/foil/PE heat sealed pouche containing double thickness sheets of additive free linear low densitiy polyethylene of 45x25 cm

Analysis Note

For more information please see:
BCR539

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service