The calculated capacity is 7.7mL. The chamber dimensions are 24mm x 40mm x 8mm.
Z741765
Convertible flow cell
Cell Only, Glass Cover Slip, sterile; γ-irradiated
Select a Size
$224.00
Estimated to ship onMarch 29, 2025
Select a Size
About This Item
$224.00
Estimated to ship onMarch 29, 2025
Recommended Products
sterility
sterile; γ-irradiated
feature
Flow Cell ONLY. No tubing included.
manufacturer/tradename
IBI CFCAS0003
General description
The detachable tops also allow the researcher to coat the glass or plastic cover slip top with a variety of materials: such as seeded cells, proteins, polymer films, and/or ECM for experiments testing the response of biofilms or cells to different materials. As with other IBI flow cells, the convertible flow cell allows the user to monitor the biofilm in real time via standard or confocal laser microscopy. Complete convertible flow cell assemblies and single cells are available, and come gamma irradiated. The devices are single use only and cannot be autoclaved for re-use.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
Already Own This Product?
-
How many mL of liquid are required to fill the chamber? (For washes, probes, etc).
1 answer-
Helpful?
-