Skip to Content
Merck
All Photos(1)

Key Documents

B1676

Millipore

Blood agar base no. 2

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$244.00

$244.00


Estimated to ship onMay 27, 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
500 G
$244.00

About This Item

UNSPSC Code:
41171605
NACRES:
NA.85

$244.00


Estimated to ship onMay 27, 2025


Request a Bulk OrderRequest more information

Agency

according to ISO 11290-1:2017
according to ISO 11290-2:2017

Quality Level

sterility

non-sterile

product line

BioChemika

form

powder

shelf life

limited shelf life, expiry date on the label

composition

Agar, 15.00 g/L
Liver extract, 2.50 g/L
Proteose peptone, 15.00 g/L
Sodium chloride, 5.00 g/L
Yeast extract, 5.00 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbe id | specific enzyme detection: suitable
microbiological culture: suitable

pH

7.4±0.2 (25 °C)

application(s)

clinical testing
environmental
food and beverages
veterinary

microbiology

suitability

Enterococcus spp.
Haemophilus spp.
Pneumococcus spp.
nonselective and differential for Campylobacter spp.
nonselective and differential for Neisseria spp.
nonselective and differential for Staphylococcus spp.
nonselective and differential for Streptococcus spp.
nonselective and differential for bacteria (General Media)
nonselective and differential for molds (General Media)
nonselective and differential for yeasts (General Media)

Application

Formulated to yield maximum recovery of fastidious pathogenic microorganisms.

Preparation Note

Suspend 21.25 g of Blood Agar Base No. 2 in 500 ml distilled water. Boil to dissolve the medium completely. Sterilize by autoclaving at 15 lbs. pressure (121°C) for 15 minutes. Cool to 40-50°C and aseptically add 7% v/v sterile defibrinated blood.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Muhammad Ahsan Anwar et al.
Microbial pathogenesis, 148, 104450-104450 (2020-08-28)
We evaluated three types of total six preparations against multidrug resistant E. coli i) three antibiotic coated ZnO nanoparticles (gentamicin coated nanoparticle-GNp; chloramphenicol coated nanoparticles-CNp; and both gentamicin & chloramphenicol coated nanoparticle-GCNp), ii) ZnO nanoparticle alone-Np, and iii) two antibiotics
Skirrow M.
British Medical Journal, 9 (1977)
A comparison of three selective media for the isolation of Brucella abortus from contaminated material.
D Hunter et al.
The British veterinary journal, 133(5), 486-489 (1977-09-01)

Articles

Chromogenic media enable the selective detection of S. aureus, which produce bluish-green colonies that are clearly differentiated from other species.

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

Streptococci- Overview of Detection, Identification, Differentiation and Cultivation Techniques

Protocols

Listeria is a public health concern, affecting elderly, pregnant, newborns, and immunocompromised individuals.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service