Skip to Content
Merck
All Photos(1)

Key Documents

74807

Millipore

Blood Free Campylobacter Medium Selective Supplement

suitable for microbiology

Synonym(s):

CCDA Selective Supplement, Campylobacter Blood-Free Medium Selective Supplement, Charcoal Cefoperazone Desoxycholate Agar Selective Supplement

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41171607
NACRES:
NA.74
Pricing and availability is not currently available.

form

powder

Quality Level

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Campylobacter spp.

General description

An antimicrobial supplement recommended for the selective isolation of Campylobacter species

Application

for the selective isolation of Campylobacter species

Components

(each vial sufficient for 500 ml medium):
Cefoperazone 16 mg

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Campylobacter spp. are regarded as the leading cause of bacterial gastroenteritis in humans worldwide.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service