Skip to Content
Merck
All Photos(1)

Documents

1.01116

Supelco

Ammonium acetate

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Ammonium acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H7NO2
CAS Number:
Molecular Weight:
77.08
MDL number:
UNSPSC Code:
12352107
EC Index Number:
211-162-9
NACRES:
NA.21

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

product line

EMSURE®

Assay

≥98.0% (acidimetric)

form

solid

impurities

≤0.005% Insoluble matter
≤0.005% Substances reducing potassium permanganate (as formic acid)
≤2% Water

ign. residue

≤0.01% (as sulfate)

pH

6.7-7.3 (25 °C, 50 g/L in H2O)

mp

114 °C

density

1.17 g/cm3 at 20 °C

bulk density

410 kg/m3

anion traces

chloride (Cl-): ≤0.0005%
nitrate (NO3-): ≤0.001%
sulfate (SO42-): ≤0.001%

cation traces

Ca: ≤0.001%
Fe: ≤0.0002%
heavy metals (as Pb): ≤0.0002%

storage temp.

15-25°C

InChI

1S/C2H4O2.H3N/c1-2(3)4;/h1H3,(H,3,4);1H3

InChI key

USFZMSVCRYTOJT-UHFFFAOYSA-N

Related Categories

Application


  • Ammonium Acetate in Buffer Preparation: A study highlighted the use of ammonium acetate in buffer solutions for molecular biology applications, emphasizing its role in maintaining pH stability and compatibility with various biochemical processes (Ahamed et al., 2024).

  • High-Purity Ammonium Acetate for Mass Spectrometry: Another research employed high-purity ammonium acetate in developing a robust mass spectrometry method for the analysis of complex biological samples, demonstrating its effectiveness in improving detection sensitivity and accuracy (Mozayad et al., 2024).

  • Material Science Applications: In the field of material science, ammonium acetate has been used as a precursor for the synthesis of novel materials with potential applications in energy storage and electronics, showcasing its versatility and functional importance (Hansen et al., 2024).

  • Pharmaceutical Synthesis: Highlighting its role in pharmaceutical formulations, a study discussed the use of ammonium acetate as a catalyst in drug synthesis processes, contributing to more efficient and cleaner production methods (Zhao et al., 2024).

  • Environmental Testing Reagent: Ammonium acetate is also pivotal in environmental science, particularly in the analysis of soil and water samples for pollution assessment, where it acts as an effective ion pairing agent to measure various pollutants (Kopeć et al., 2024).


Linkage

Replaces: AX1220; AX1220-1; AX1220-3; AX1220-5; AX1220-20

Analysis Note

Assay (acidimetric): ≥ 98.0 %
Insoluble matter: ≤ 0.005 %
pH-value (5 %; water, 25 °C): 6.7 - 7.3
Chloride (Cl): ≤ 0.0005 %
Nitrate (NO₃): ≤ 0.001 %
Sulfate (SO₄): ≤ 0.001 %
Heavy metals (as Pb): ≤ 0.0002 %
Ca (Calcium): ≤ 0.001 %
Fe (Iron): ≤ 0.0002 %
Substances reducing potassium permanganate (as formic acid): ≤ 0.005 %
Residue on ignition (as sulfate): ≤ 0.01 %
Water: ≤ 2.0 %
Corresponds to ACS,Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

EMSURE® Grade Inorganic Solvents and Reagents offer exceptional purity and quality control, ensuring reliable, compliant results for regulated analyses.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service