Skip to Content
Merck
All Photos(2)

Key Documents

92332

Supelco

Chloride ionophore IV

Selectophore, for ion-selective electrodes

Synonym(s):

4,5-Bis-[N′-(butyl)thioureido]-2,7-di-tert-butyl-9,9-dimethylxanthene

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$1,170.00

$1,170.00


Estimated to ship onMay 27, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$1,170.00

About This Item

Empirical Formula (Hill Notation):
C33H50N4OS2
CAS Number:
Molecular Weight:
582.91
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

$1,170.00


Estimated to ship onMay 27, 2025


Request a Bulk Order

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

form

powder

SMILES string

CCCCNC(=S)Nc1cc(cc2c1Oc3c(NC(=S)NCCCC)cc(cc3C2(C)C)C(C)(C)C)C(C)(C)C

InChI

1S/C33H50N4OS2/c1-11-13-15-34-29(39)36-25-19-21(31(3,4)5)17-23-27(25)38-28-24(33(23,9)10)18-22(32(6,7)8)20-26(28)37-30(40)35-16-14-12-2/h17-20H,11-16H2,1-10H3,(H2,34,36,39)(H2,35,37,40)

InChI key

GYGZJHGSLBVTPV-UHFFFAOYSA-N

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Wenjun Di et al.
Analytical methods : advancing methods and applications, 12(11), 1441-1448 (2020-04-01)
Personalized approaches for continuous monitoring of chloride levels are potentially valuable for evaluating the efficacy of new treatments of genetic disorders such as cystic fibrosis. In this report, we validated optode-based nanosensors for real-time chloride monitoring in the interstitial fluid
Chloride ionophore, detection limit ~0.0000065 M chloride. Detection of chloride in biological samples.
K.P. Xiao et al.
Analytical Chemistry, 69, 1038-1038 (1997)
Strong Hydrogen Bond-Mediated Complexation of H2PO4- by Neutral Bis-Thiourea Hosts
P. Buhlmann et al.
Tetrahedron, 53, 1647-1647 (1997)
Antra Ganguly et al.
SLAS technology, 25(1), 25-32 (2019-10-17)
Misclassification of an acute disease condition as chronic and vice versa by electrochemical sweat biomarker sensors can cause significant psychological, emotional, and financial stress among patients. To achieve higher accuracy in distinguishing between a chronic condition and an acute condition

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service