Skip to Content
Merck
All Photos(2)

Key Documents

24894

Supelco

Chloride ionophore III

Selectophore

Synonym(s):

3,6-Didodecyloxy-4,5-dimethyl-o-phenylene-bis(mercury chloride), ETH 9033

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$693.00

$693.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
50 MG
$693.00

About This Item

Empirical Formula (Hill Notation):
C32H56Cl2Hg2O2
CAS Number:
Molecular Weight:
944.87
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

$693.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

product line

Selectophore

Quality Level

SMILES string

CCCCCCCCCCCCOc1c(C)c([Hg]Cl)c([Hg]Cl)c(C)c1OCCCCCCCCCCCC

InChI

1S/C32H56O2.2ClH.2Hg/c1-5-7-9-11-13-15-17-19-21-23-27-33-31-29(3)25-26-30(4)32(31)34-28-24-22-20-18-16-14-12-10-8-6-2;;;;/h5-24,27-28H2,1-4H3;2*1H;;/q;;;2*+1/p-2

InChI key

SSQLFLVGBOQWGG-UHFFFAOYSA-L

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples
ETH 9033 (chromoionophore III) may be used to monitor Cl- activity during the development of chloride sensitive probes encapsulated by biologically localized embedding.[1] It may also be used with Cl- ion sensitive ISFETs (ion-sensitive field-effect transistor) with photocured polyurethane-based polymer membrane to study sodium chloride solutions and to determine the chloride ions.[2]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Chloride selective ionophore for Cl--electrodes[3]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Murphy G Brasuel et al.
The Analyst, 128(10), 1262-1267 (2003-12-12)
The first nanometer scale anion sensing fluorescent spherical nanosensors, or PEBBLEs (probes encapsulated by biologically localized embedding) have been developed for the intracellular monitoring of chloride. The general scheme for the polymerization and introduction of sensing components creates a matrix
M. Rothmaier et al.
Analytica Chimica Acta, 327, 17-17 (1996)
Investigation of chloride sensitive ISFETs with different membrane compositions suitable for medical applications.
Bratov, Andrey, Natalia Abramova, and Carlos Domi'nguez.
Analytica Chimica Acta, 514, 99-106 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service