Skip to Content
Merck
All Photos(2)

Key Documents

71747

Supelco

Sodium ionophore X

for ion-selective electrodes, Selectophore, function tested

Synonym(s):

4-tert-Butylcalix[4]arene-tetraacetic acid tetraethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$270.00

$270.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$270.00

About This Item

Empirical Formula (Hill Notation):
C60H80O12
CAS Number:
Molecular Weight:
993.27
Beilstein:
3587002
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

$270.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

form

crystals

quality

function tested

SMILES string

CCOC(=O)COc1c2Cc3cc(cc(Cc4cc(cc(Cc5cc(cc(Cc1cc(c2)C(C)(C)C)c5OCC(=O)OCC)C(C)(C)C)c4OCC(=O)OCC)C(C)(C)C)c3OCC(=O)OCC)C(C)(C)C

InChI

1S/C60H80O12/c1-17-65-49(61)33-69-53-37-21-39-27-46(58(8,9)10)29-41(54(39)70-34-50(62)66-18-2)23-43-31-48(60(14,15)16)32-44(56(43)72-36-52(64)68-20-4)24-42-30-47(59(11,12)13)28-40(55(42)71-35-51(63)67-19-3)22-38(53)26-45(25-37)57(5,6)7/h25-32H,17-24,33-36H2,1-16H3

InChI key

HZHADWCIBZZJNV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium ionophore X is generally used in the preparation of ion-selective electrode (ISE).
Visit our Sensor Applications portal to learn more.

Application

Selected application examples
It may be used in a comparison study of ISE-based electronic tongue for direct and two-stage data analysis on Artificial Neural Network (ANN), Partial Least Squares-Discriminant Analysis (PLS-DA)) and Principal Components Analysis (PCA).[1]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Ionophore employed in solid-state and polymeric membrane sodium-selective electrodes[2][3]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analytical Chemistry, 64, 2496-2496 (1992)
D. Diamond et al.
Analyst, 114, 1551-1551 (1989)
Direct and two-stage data analysis procedures based on PCA, PLS-DA and ANN for ISE-based electronic tongue-Effect of supervised feature extraction.
Ciosek P
Talanta, 67(3), 590-596 (2005)
Yihang Chen et al.
Lab on a chip, 20(22), 4205-4214 (2020-10-14)
Microchannels in hydrogels play an essential role in enabling a smart contact lens. However, microchannels have rarely been created in commercial hydrogel contact lenses due to their sensitivity to conventional microfabrication techniques. Here, we report the fabrication of microchannels in

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service